![Minimum Essential Medium Eagle HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/126/390/c6e67f91-432c-4b53-81c5-2dc349513ee9/640/c6e67f91-432c-4b53-81c5-2dc349513ee9.jpg)
Minimum Essential Medium Eagle
![Minimum Essential Medium Eagle HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/126/390/c6e67f91-432c-4b53-81c5-2dc349513ee9/640/c6e67f91-432c-4b53-81c5-2dc349513ee9.jpg)
Synonym(s):
MEM, Minimum Essential Medium Eagle
Products
Product Category
Brand
Physical Form
Earles Salts
Glucose (g/L)
Hanks Salts
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-20 of 31
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2