Skip to Content
Merck
All Photos(1)

Key Documents

L9529

Sigma-Aldrich

Latex beads, amine-modified polystyrene, fluorescent orange

aqueous suspension, 2.0 μm mean particle size

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$104.00

$104.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
1 ML
$104.00

About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.56

$104.00


Estimated to ship onMay 29, 2025


form

aqueous suspension

composition

Solids, 2.4-2.6%

mean particle size

2.0 μm

fluorescence

λex ~520 nm; λem ~540 nm

Looking for similar products? Visit Product Comparison Guide

Application

Latex beads, amine-modified polystyrene, fluorescent orange has been used in functionalized microbeads for rapid antimicrobial susceptibility testing (AST) evaluation.[1][2] It has also been used in microparticle functionalization for the detection of live bacteria.[3]

Biochem/physiol Actions

Amine-modified polystyrene latex beads have been used to develop an electrochemical nitrite nanosensor as well as to validate a pharyngeal aspiration technique for exposing the mouse lung to respirable particles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Geys et al.
Toxicology letters, 160(3), 218-226 (2005-09-03)
Recent studies indicate that inhaled ultrafine particles can pass into the circulation. To study this translocation in an in vitro model three types of pulmonary epithelial cells were examined. The integrity of the cell monolayer was verified by measuring the
Development of a self-driving bioassay based on diffusion for simple detection of microorganisms
Wang J C, et al.
Sensors and Actuators B, Chemical, 278, 140-146 (2019)
Ninad Mehendale et al.
Biomedical microdevices, 20(1), 6-6 (2017-12-01)
Pillar-based microfluidic sorting devices are preferred for isolation of rare cells due to their simple designs and passive operation. Dead-end pillar filters can efficiently capture large rare cells, such as, circulating tumor cells (CTCs), nucleated red blood cells (NRBCs), CD4
Jhih-Cheng Wang et al.
ACS sensors, 3(10), 2182-2190 (2018-09-18)
Rapid detection of bacteria and their susceptibility to specific antibiotics plays a vital role in microbial infection treatments. Antimicrobial susceptibility testing (AST) is a common measure to select effective drugs. However, the conventional practices, such as broth dilution, E-test, and
G V S Rao et al.
Journal of toxicology and environmental health. Part A, 66(15), 1441-1452 (2003-07-15)
Recent studies have demonstrated that the mouse lung can be exposed to soluble antigens by aspiration of these antigens from the pharynx. This simple technique avoids the trauma associated with intratracheal instillation. In this study, the pharyngeal aspiration technique was

Questions

  1. The specified output emission is 540nm, which is green. However, the beads are labelled "yellow to orange". When exciting with green I detect fluorescence at 600nm on my spectrometer, which is yellow to orange. Is 540 the actual expected emission? or 600?

    1 answer
    1. The excitation range for this product is 470 nm to 525 nm, and the emission range is 535 nm to 650 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service