Skip to Content
Merck
All Photos(1)

Key Documents

L6649

Sigma-Aldrich

Lonomycin A sodium salt from Streptomyces ribosidificus

90-95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H75NaO14
CAS Number:
Molecular Weight:
851.05
Beilstein:
4122659
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

90-95% (TLC)

storage temp.

2-8°C

SMILES string

[Na+].CO[C@H]1C[C@]2(CC[C@](C)(O2)[C@H]3CC[C@](C)(O3)[C@@H]4O[C@H]([C@H](OC)[C@@H]4C)[C@H]5O[C@](C)(O)[C@H](C)[C@@H](OC)[C@@H]5C)O[C@@H]([C@@H]1C)[C@@H](C)[C@H]6O[C@](O)([C@H](C)[C@@H](OC)[C@H]6C)[C@H](C)C([O-])=O

InChI

1S/C44H76O14.Na/c1-21-29(49-12)20-43(56-31(21)22(2)32-23(3)34(51-14)27(7)44(48,57-32)28(8)39(45)46)19-18-40(9,58-43)30-16-17-41(10,54-30)38-25(5)35(52-15)37(53-38)36-24(4)33(50-13)26(6)42(11,47)55-36;/h21-38,47-48H,16-20H2,1-15H3,(H,45,46);/q;+1/p-1/t21-,22-,23+,24+,25+,26-,27-,28-,29+,30-,31+,32-,33+,34+,35-,36+,37-,38-,40+,41+,42+,43-,44-;/m1./s1

InChI key

PIKGYGHAIOUQMF-SZCYNZSUSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Application of nuclear magnetic resonance to bio-organic chemistry].
J Y Lallemand
Bulletin et memoires de l'Academie royale de medecine de Belgique, 137(8-9), 453-456 (1982-01-01)
B Marks et al.
Current biology : CB, 8(13), 740-749 (1998-07-04)
Following exocytosis at the synapse, synaptic vesicle components are recovered by endocytosis. Morphological analysis has suggested that this occurs by a clathrin-mediated pathway, and the GTPase dynamin is thought to be involved in 'pinching off' endocytosing vesicles. The finding that
A Saito et al.
Zentralblatt fur Bakteriologie, Mikrobiologie, und Hygiene. Series A, Medical microbiology, infectious diseases, virology, parasitology, 264(3-4), 446-454 (1987-05-01)
The inhibition of Toxoplasma multiplication inside cells does not correlate with an enhanced release of oxygen intermediates except in the case of peritoneal macrophages treated with Obioactin. The inhibition observed in alveolar macrophages treated with Obioactin, in kidney cells treated
D S Hoover et al.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research, 8(12), 1371-1380 (1998-01-07)
Expression of Pim-1, an oncogenic serine/threonine kinase, is highly regulated at the transcriptional, posttranscriptional, and posttranslational levels. Here, we report that expression of Pim-1 kinase is additionally regulated at the translational level. Pim-1 protein expression did not increase in Hut-78
T Mizutani et al.
The Journal of antibiotics, 33(11), 1224-1230 (1980-11-01)
Lonomycin B (II), C44H75O14Na, m.p. 181-182 degrees C, and lonomycin C (III), C43H73O14Na, m.p 186-187 degrees C, were isolated as their sodium salts from the fermentation broth of Streptomyces ribosidificus TM-481. Their physicochemical properties demonstrated that II and III are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service