Key Documents
L6164
Laser Tough-Spots®
diam. 1/2 in., green
Synonym(s):
solvent resistant labels, temperature resistant labels
Select a Size
1 PKG
$188.00
$188.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 PKG
$188.00
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
$188.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
material
green
polyolefin
packaging
pack of 3840 ea (192 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-2200
diam.
1/2 in.
Related Categories
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)