Skip to Content
Merck
All Photos(1)

Key Documents

Z743582

Laser Tough-Spots®

diam. 1/2 in., white, pack of 3840 (192 labels per sheet)

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing

Select a Size

3840 EA
$158.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
3840 EA
$158.00

About This Item

UNSPSC Code:
41103032
NACRES:
NB.35

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 3840 (192 labels per sheet)

manufacturer/tradename

Diversified Biotech SPOT-2000

diam.

1/2 in.

color

white

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.

Legal Information

Tough-Spots is a registered trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can these withstand liquid nitrogen? 

    1 answer
    1. Yes, this product can withstand liquid nitrogen.
      Please see the link below for more information:
      https://www.divbio.com/product/spot-2000

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service