Skip to Content
Merck
All Photos(1)

Key Documents

L6006

Sigma-Aldrich

Leu-Pro trifluoroacetate salt

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O3 · C2HF3O2
CAS Number:
Molecular Weight:
342.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

powder

color

white

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](N)C(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C11H20N2O3.C2HF3O2/c1-7(2)6-8(12)10(14)13-5-3-4-9(13)11(15)16;3-2(4,5)1(6)7/h7-9H,3-6,12H2,1-2H3,(H,15,16);(H,6,7)/t8-,9-;/m0./s1

InChI key

AUVSSRGHMVVWPD-OZZZDHQUSA-N

Biochem/physiol Actions

L-leucyl-L-proline (Leu-Pro) and its retro variant Pro-Leu may be used along with other proline dipeptides to study imprinting and chemotaxis in tetrahymena.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengxia Zhang et al.
Molecular bioSystems, 6(5), 852-861 (2010-06-23)
A novel metabonomic method based on fast liquid chromatography coupled with ion trap-time of flight mass spectrometry (UFLC/MS-IT-TOF) was applied to study the metabolic changes of plasma and urine in depression and excess fatigue rats. Principal component analysis (PCA) and
G Csaba et al.
Bioscience reports, 17(6), 537-542 (1998-04-30)
Proline-glycine, proline-leucine and proline-valine dipeptides and their retro variants were used in the experiments to study the effects of pretreatment (imprinting) in Tetrahymena, by investigating fluorescein isothiocyanate (FITC)-conjugated peptide binding. The protozoan organism could differentiate between the proline-dipeptides containing different
L Köhidai et al.
Cell biology international, 21(6), 341-345 (1997-06-01)
Our investigations demonstrate that proline-containing dipeptides can provoke a chemosensory response from the unicellular Tetrahymena pyriformis. The chemotactic effects of the dipeptides have a close relationship with the side chain and the lipophilicity of the amino-terminal amino acid. Comparison of
G Schoetz et al.
Electrophoresis, 22(12), 2409-2415 (2001-08-25)
Dynamic capillary electrophoresis (DCE) and computer simulation of the elution profiles with the theoretical plate and the stochastic model has been applied to determine the isomerization barriers of the three dipeptides L-alanyl-L-proline, L-leucyl-L-proline, and L-phenylalanyl-L-proline. The separation of the rotational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service