Skip to Content
Merck
All Photos(1)

Key Documents

L5631

Sigma-Aldrich

L-Lysine–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

form

saline suspension

extent of labeling

5-15 μmol per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

L-lysine-agarose is used in protein chromatography, affinity chromatography and amino acid resins. L-lysine-agarose has been used to study mitogen-activated protein kinase (MAPK) cascades in abscisic acid (ABA) signal transduction pathways as well as to study the regulation of phosphorylation of tau protein in the brain.

Physical form

Suspension in 2.0 M NaCl containing preservative

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O D Ratnoff et al.
Blood, 67(6), 1550-1553 (1986-06-01)
Purification of Hageman factor (HF, factor XII) from human plasma is a tedious procedure and the product is not always in the precursor form. Hojima has described a protein derived from corn kernels that inhibits the enzymatic properties of HF.
Complement C5b-9 Increases Plasminogen Binding and Activation on Human Endothelial Cells.
Christiansen, V.J., et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 16, 164-171 (1997)
Predominant expression of the ? subunit of prolyl 4-hydroxylase (disulfide isomerase) in human extravillous trophoblasts.
Vanderpuye, O.A., et al.
Histochemistry and Cell Biology, 100(3), 241-246 (1993)
J L Brash et al.
Blood, 71(4), 932-939 (1988-04-01)
The transient detection of fibrinogen on surfaces has been described (Vroman effect) and high-mol-wt kininogen (HK) has been shown to play a role in this reaction. In this study, we attempted to identify the form of HK responsible for preventing
Piotr Borsuk et al.
Biological chemistry, 388(2), 135-144 (2007-01-31)
Expression of the arginase structural gene (agaA) in Aspergillus nidulans is subject to complex transcriptional and post-transcriptional regulation. Arginase mRNA has a long 5'-UTR sequence. Analysis of this sequence in silico revealed its putative complex secondary structure, the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service