Skip to Content
Merck
All Photos(1)

Key Documents

B8312

Sigma-Aldrich

BNTX maleate salt hydrate

≥98% (HPLC)

Synonym(s):

7(E)-Benzylidenenaltrexone maleate salt hydrate, BNTX-7 maleate salt hydrate, Morphinan-6-one,17-(cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxy-7-(phenylmethylene)-,(5a,7E)- maleate salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$171.00
25 MG
$636.00

$171.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$171.00
25 MG
$636.00

About This Item

Empirical Formula (Hill Notation):
C31H31NO8 · xH2O
Molecular Weight:
545.58 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$171.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

O.OC(=O)\C=C/C(O)=O.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)\C(C[C@@]35O)=C\c6ccccc6)CC7CC7

InChI

1S/C27H27NO4.C4H4O4.H2O/c29-20-9-8-18-13-21-27(31)14-19(12-16-4-2-1-3-5-16)23(30)25-26(27,22(18)24(20)32-25)10-11-28(21)15-17-6-7-17;5-3(6)1-2-4(7)8;/h1-5,8-9,12,17,21,25,29,31H,6-7,10-11,13-15H2;1-2H,(H,5,6)(H,7,8);1H2/b19-12+;2-1-;/t21-,25+,26+,27-;;/m1../s1

InChI key

AGUNLJBLOJAUKM-ZKJXOIBSSA-N

Application

BNTX, a selective nonpeptide σ1 opioid receptor (DOR1) antagonist, is used in σ opioid receptor research to help differentiate and characterized the function and role of the σ1 opioid receptor versus other σ opioid receptor subtypes.[1] BNTX was used to help define the role of σ1 opioid receptors in neurological processes such as anxiety. It was used to help define the role of σ opioid receptors in myocardial infarct development and size.[2][3]

Biochem/physiol Actions

BNTX is a selective d1 non peptide opioid receptor antagonist.
BNTX is a selective nonpeptide δ1 opioid receptor antagonist. It antagonizes the effect of D-Pen2,5-enkephalin (a δ1 agonist 4-6 fold, but did not affect the activity of δ2 or μ agonists.<<<107>>> BNTX is used to discriminate among opioid receptor subtypes in effects such as alcohol and drug dependence. It binds 100× more tightly to δ1 than to δ2 receptors. In the tail-flick assay, antinocieceptive ED50 = 646.4 pmol/mouse.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshinori Miyata et al.
Bioorganic & medicinal chemistry letters, 21(16), 4710-4712 (2011-07-19)
We evaluated antimalarial and/or chloroquine-resistance reversing effects of five opioid receptor antagonists. Although none of the evaluated compounds showed antimalarial effects, some of them, especially the δ(1) receptor antagonist, 7-benzylidenenaltrexone (BNTX) exhibited potent chloroquine-resistance reversing effects in Plasmodium chabaudi.
Garrett J Gross et al.
Journal of cardiovascular pharmacology, 59(2), 194-197 (2011-12-02)
Eribis peptide 94 (EP 94) is a novel enkephalin derivative that binds with high potency to μ and δ opioid receptors with less affinity for the κ opioid receptor. This compound has recently been shown to produce an acute reduction
Azusa Sugiyama et al.
Neuropharmacology, 79, 314-320 (2013-12-18)
Recently, we reported that the δ opioid receptor (DOR) agonist KNT-127 produces anxiolytic-like effects in behaving rats. Here, we report on the roles of DOR subtypes ( DOR(1) and DOR(2)) play in mediating KNT-127-induced anxiolytic-like effects. Pretreatment with the DOR(2)-selective
So Young Kim et al.
Oncotarget, 8(37), 61057-61071 (2017-10-06)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a potential biological anticancer agent. However, a wide range of human primary cancers, including pancreatic cancer, display resistance to apoptosis induction by TRAIL. Therefore, this resistance needs to be overcome to allow TRAIL

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service