Skip to Content
Merck
All Photos(1)

Documents

L2898

Sigma-Aldrich

Lysostaphin from Staphylococcus staphylolyticus

aseptically filled

Synonym(s):

Glycyl-glycine Endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

sterility

aseptically filled

Quality Level

form

powder

specific activity

>500 units/mg protein

mol wt

25 kDa

composition

Protein, 40-70% biuret

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide
Lysostaphin is mainly liberated as a proenzyme. It is highly stable at pH 4 and temperature 5°C.

Biochem/physiol Actions

Lysostaphin exhibit a lytic action against Staphylococcus aureus. It possesses several functions of three major enzymes, such as, glycylglycine endopeptidase, endo-β-N-acetyl glucosamidase and N-acteyl muramyl-L-alanine amidase. Lysostaphin can be used to treat antibiotic-resistant staphylococcal infections. Lysostaphin being an antistaphylococcal agent, can be used as a preservative in the food industry and in clinical labs for rapid screening.
Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. Because lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
pH Optimum for activity: ~7.5

Unit Definition

One unit will reduce the turbidity (A620) of a suspension of Staphylococcus aureus cells from 0.250 to 0.125 in 10 min at pH 7.5 at 37 °C in a 6.0 ml reaction mixture.

Preparation Note

Prepared from L 7386

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysostaphin: an antistaphylococcal agent
Kumar J K
Applied Microbiology and Biotechnology, 80(4), 555-561 (2008)
Thomas S Lawson et al.
Journal of clinical laboratory analysis, 25(5), 359-365 (2011-09-16)
Aspects of the fluorescence in situ hybridization (FISH) method for the detection of clinically important bacteria, such as Staphylococcus aureus, Staphylococcus epidermidis, and Escherichia coli, were investigated for optimization. Various approaches to optimizing the FISH procedure were taken and different
R Satishkumar et al.
Nanotechnology, 22(50), 505103-505103 (2011-11-24)
The objective of this paper was to study the effect of antibody-directed targeting of S. aureus by comparing the activities of lysostaphin conjugated to biodegradable polylactide nanoparticles (NPs) in the presence and in the absence of co-immobilized anti-S. aureus antibody.
Yuliya Yurko et al.
Surgical innovation, 19(1), 20-26 (2011-07-12)
Naturally occurring antimicrobial peptides are possibly the "next frontier" in infection prevention. Binding them to mesh could reduce the rate of mesh infections. This study identifies an antimicrobial agent capable of significant antibacterial activity when bound to mesh. Lysozyme, human
Igor Belyansky et al.
The American surgeon, 77(8), 1025-1031 (2011-09-29)
Mesh and wound infections during hernia repair are predominantly caused by Staphylococcus aureus. Human acellular dermis (HAD) is known to lose its integrity in the face of large bacterial loads. The goal of this study was to determine if lysostaphin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service