Skip to Content
Merck
All Photos(2)

Key Documents

SRE0007

Sigma-Aldrich

Mutanolysin from Streptomyces globisporus ATCC 21553

Suitable for manufacturing of diagnostic kits and reagents, ≥4000 units/mg protein (biuret)

Synonym(s):

Mutanolysin

Sign Into View Organizational & Contract Pricing

Select a Size

25000 UNITS
$740.00
100000 UNITS
$1,970.00

$740.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
25000 UNITS
$740.00
100000 UNITS
$1,970.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$740.00


Estimated to ship onMay 30, 2025


biological source

bacterial (Streptomyces globisporus)

Quality Level

form

lyophilized powder

specific activity

≥4000 units/mg protein (biuret)

mol wt

23 kDa

concentration

≥40% protein

suitability

suitable for cell lysis

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Mutanolysin from Streptomyces globisporus consists of two main lytic enzymes and may be a useful agent for dental caries control

Application

Provides gentle cell lysis for the isolation of easily degradable biomolecules and RNA from bacteria. It has been used in the formation of spheroplasts for isolation of DNA.

Biochem/physiol Actions

Mutanolysin cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. It lyses Listeria and other gram positive bacteria such as Lactobacillus and Lactococcus.
Mutanolysin is an N-acetylmuramidase. Like lysozyme, it is a muralytic enzyme that cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. Its carboxy terminal moieties are involved in the recognition and binding of unique cell wall polymers. Mutanolysin lyses Listeria and other Gram-positive bacteria such as Lactobacillus and Lactococcus.

Unit Definition

One unit will produce a ΔA600 of 0.01 per minute at pH 6.0 at 37 °C in a 1 mL volume using a suspension of Streptococcus faecalis cell wall as substrate.

Physical form

Lyophilized powder containing Ficoll® and sodium succinate buffer salts

Legal Information

Ficoll is a registered trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 本製品に含まれる3種類の酵素 1. N-acetylmuramidase M1 2. N-acetylmuramidase M2 3. N-Acetylmuramyl-L-alanine amidase それぞれのアミノ酸配列は同定されていますか。 また、配列を開示いただくことは可能でしょうか。

    1 answer
    1. Unfortunately, this material has not been sequenced.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service