Skip to Content
Merck
All Photos(1)

Documents

L1500

Sigma-Aldrich

DL-Lactic acid lithium salt

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O3Li
CAS Number:
Molecular Weight:
96.01
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

~98%

SMILES string

[Li+].CC(O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1

InChI key

GKQWYZBANWAFMQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains approximately equal amounts of DL isomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Gabba et al.
Biophysical journal, 118(2), 422-434 (2019-12-18)
We present a fluorescence-based approach for determination of the permeability of small molecules across the membranes of lipid vesicles and living cells. With properly designed experiments, the method allows us to assess the membrane physical properties both in vitro and in vivo.
E Brindha et al.
Toxicology mechanisms and methods, 25(2), 150-154 (2015-01-07)
This study was aimed to evaluate the preventive role of phytic acid on lysosomal enzymes in isoproterenol (ISO)-induced myocardial infarction (MI) in male Wistar rats. Rats subcutaneously injected with ISO (85 mg/kg) at an interval of 24 h for two days showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service