Skip to Content
Merck
All Photos(1)

Key Documents

L0649

Sigma-Aldrich

Leupeptin hydrochloride

powder, ≥70% (HPLC)

Synonym(s):

Acetyl-Leu-Leu-Arg-al hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$119.00
25 MG
$417.00
100 MG
$1,380.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$119.00
25 MG
$417.00
100 MG
$1,380.00

About This Item

Empirical Formula (Hill Notation):
C20H38N6O4 · HCl
CAS Number:
Molecular Weight:
463.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Product Name

Leupeptin hydrochloride, microbial, ≥70% (HPLC)

biological source

microbial

Quality Level

Assay

≥70% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to yellow

storage temp.

−20°C

SMILES string

Cl.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCNC(N)=N)C=O

InChI

1S/C20H38N6O4.ClH/c1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;/h11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);1H

InChI key

OQQYPQNFLLAILR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM.
Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Analysis Note

Leupeptin gives multiple peaks on HPLC because of the formation of tautomeric isomers in solution. Majority of contaminating peptide is racemized leupeptin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arne Fabritius et al.
Cell chemical biology, 25(12), 1554-1561 (2018-09-18)
Protein engineering involves generating and screening large numbers of variants for desired properties. While modern DNA technology has made it easy to create protein diversity on the DNA level, the selection and validation of candidate proteins from large libraries remains
Determination of ERK activity: antiphospho-ERK antibodies, in vitro phosphorylation, and in-gel kinase assay.
Sarah Kraus et al.
Methods in molecular biology (Clifton, N.J.), 250, 29-48 (2004-02-03)
Suzanne Elsasser et al.
Methods in molecular biology (Clifton, N.J.), 832, 403-422 (2012-02-22)
The proteasome is an ATP-dependent molecular machine that degrades proteins through the concerted activity of dozens of subunits. It is the yin to the ribosome's yang, and together these entities mold the protein landscape of the cell. Native gels are
Michaela Nejedla et al.
Molecular biology of the cell, 27(15), 2381-2393 (2016-06-17)
Profilin controls actin nucleation and assembly processes in eukaryotic cells. Actin nucleation and elongation promoting factors (NEPFs) such as Ena/VASP, formins, and WASP-family proteins recruit profilin:actin for filament formation. Some of these are found to be microtubule associated, making actin
Shanti Pal Gangwar et al.
STAR protocols, 2(4), 100855-100855 (2021-10-15)
Ionotropic glutamate receptors (iGluRs) are ligand-gated ion channels that play crucial roles in the central nervous system. iGluR homologs, termed glutamate receptor-like channels (GLRs), have been found in plants. Investigating the structural and functional relationship between iGluRs and GLRs was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service