Merck
  • Purification and cryo-EM structure determination of Arabidopsis thaliana GLR3.4.
Purification and cryo-EM structure determination of Arabidopsis thaliana GLR3.4.
STAR protocols (2021-10-15)
Shanti Pal Gangwar, Marriah N Green, Maria V Yelshanskaya, Alexander I Sobolevsky
ABSTRACT
Ionotropic glutamate receptors (iGluRs) are ligand-gated ion channels that play crucial roles in the central nervous system. iGluR homologs, termed glutamate receptor-like channels (GLRs), have been found in plants. Investigating the structural and functional relationship between iGluRs and GLRs was limited by GLR protein expression, purification, and structural characterization. Here, we provide a detailed protocol for Arabidopsis thaliana GLR3.4 (AtGLR3.4) expression in a mammalian cell line and purification for structure determination by cryogenic electron microscopy (cryo-EM). For the complete details on the use and execution of this protocol, please refer to Green et al. (2021).
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥70% (HPLC)
Sigma-Aldrich
d-Desthiobiotin, ≥98% (TLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥75% (HPLC)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
Sigma-Aldrich
L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)