Skip to Content
Merck
All Photos(1)

Key Documents

K3519

Sigma-Aldrich

KRM-III

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1,4-diphenyl-2H-imidazole-2-thione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$142.00
25 MG
$749.00

$142.00


Estimated to ship onMarch 21, 2025



Select a Size

Change View
5 MG
$142.00
25 MG
$749.00

About This Item

Empirical Formula (Hill Notation):
C15H12N2S
CAS Number:
Molecular Weight:
252.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$142.00


Estimated to ship onMarch 21, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to light tan

solubility

DMSO: soluble ≥20 mg/mL

storage temp.

2-8°C

SMILES string

S=C1NC(=CN1c2ccccc2)c3ccccc3

InChI

1S/C15H12N2S/c18-15-16-14(12-7-3-1-4-8-12)11-17(15)13-9-5-2-6-10-13/h1-11H,(H,16,18)

InChI key

QKOAQVKXZJFMET-UHFFFAOYSA-N

Biochem/physiol Actions

KRM-III is an inhibitor of T cell antigen receptor (TCR), an inhibitor of phorbol myristate acetate (PMA) /ionomycin-induced nuclear factor of activated T cells (NFAT) activation and T cell proliferation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service