Skip to Content
Merck
All Photos(3)

Key Documents

I5127

Sigma-Aldrich

2′,3′-O-Isopropylideneuridine

≥99% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$464.00
25 G
$1,790.00

$464.00


Estimated to ship onMay 17, 2025



Select a Size

Change View
5 G
$464.00
25 G
$1,790.00

About This Item

Empirical Formula (Hill Notation):
C12H16N2O6
CAS Number:
Molecular Weight:
284.27
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

$464.00


Estimated to ship onMay 17, 2025


biological source

synthetic (organic)

Quality Level

Assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N3C=CC(=O)NC3=O

InChI

1S/C12H16N2O6/c1-12(2)19-8-6(5-15)18-10(9(8)20-12)14-4-3-7(16)13-11(14)17/h3-4,6,8-10,15H,5H2,1-2H3,(H,13,16,17)/t6-,8-,9-,10-/m1/s1

InChI key

GFDUSNQQMOENLR-PEBGCTIMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′,3′-O-Isopropylideneuridine is used in the chemical synthesis of N-benzoylated uridine derivatives and N3-substituted 2′,3′-O-isopropylideneuridines with central nervous system (CNS) depressant activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomomi Shimizu et al.
Chemical & pharmaceutical bulletin, 53(3), 313-318 (2005-03-04)
Seventy eight N(3)-substituted derivatives of uridine (1), thymidine (2), 2'-deoxyuridine (3), 6-azauridine (4), 2',3'-O-isopropylideneuridine (5), and arabinofuranosyluracil (6) were synthesized and their antinociceptive effects were evaluated. N(3)-(2',4'-Dimethoxyphenacyl)uridine (1l), N(3)-(2',4'-dimethoxyphenacyl)2'-deoxyuridine (3l), and N(3)-(2',5'-dimethoxyphenacyl)arabinofuranosyluracil (6m) possessed 93, 86, and 82% of the
C S Yao et al.
Chemical & pharmaceutical bulletin, 47(12), 1802-1804 (2000-04-05)
N3-Substituted derivatives of 2',3'-O-isopropylideneuridine (1) were synthesized and their pharmacological effects on the central nervous system (CNS) examined using mice. Methyl (2), ethyl (3), propyl (4), butyl (5), allyl (6), benzyl (7), o-, m-, p-xylyls (8, 9, 10), and alpha-phenylethyl
M L Scholes et al.
International journal of radiation biology, 61(4), 443-449 (1992-04-01)
The effect of pH on base release in the gamma-radiolysis of N2O-saturated solutions of a number of nucleosides (including uridine, 3-methyluridine, 2',3'-O-isopropylidene-uridine, and adenosine) has been investigated. For all these nucleotides, independent of the base or sugar moiety, base release
S I Malekin et al.
Bioorganicheskaia khimiia, 23(8), 648-654 (1997-08-01)
New nucleoside-phospholipid conjugates were synthesized based on 1,2-disubstituted glycerides and nucleosides. These contain rac-1-hexadecyl-2-palmitoyl(or 2-methylcarbamoyl)-sn-glycero-3-phosphate as the phospholipid component and 2',3'-didehydro-3'-deoxythymidine, 1-(Z-5-hydroxypentene-2-yl)thymine, or 2',3'-isopropylideneuridine as a nucleoside component. The conjugates were synthesized by three different ways: from rac-1-hexadecyl-2-acyl-sn-glycero-3-phospodichlorides, -3-phosphatidic acids
Anita R Maguire et al.
Carbohydrate research, 337(4), 369-372 (2002-02-14)
An improved procedure for the synthesis of N-benzoyl-2',3'-O-isopropylidene uridine via one-step selective N-benzoylation of 2',3' -O-isopropylidene uridine has been developed. An efficient synthetic route to N-benzoyl thymidine via initial tribenzoylation, followed by selective hydrolysis of the benzoates is also described.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service