Skip to Content
Merck
All Photos(1)

Key Documents

I9766

Sigma-Aldrich

D-myo-Inositol 1,4,5-tris-phosphate trisodium salt

≥95% (by 1H NMR and TLC)

Synonym(s):

Ins(1,4,5)P3

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$190.00
1 MG
$254.00

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
$190.00
1 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C6H12O15P3Na3
CAS Number:
Molecular Weight:
486.04
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (by 1H NMR and TLC)

form

powder

color

white

solubility

H2O: soluble

cation traces

K: <0.2%
Na: 12.5-16.0

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[P](=O)([O-])(O[C@H]1[C@@H]([C@H]([C@H]([C@H]([C@@H]1O)O[P](=O)([O-])O)O)O)O[P](=O)([O-])O)O

InChI

1S/C6H15O15P3.3Na/c7-1-2(8)5(20-23(13,14)15)6(21-24(16,17)18)3(9)4(1)19-22(10,11)12;;;/h1-9H,(H2,10,11,12)(H2,13,14,15)(H2,16,17,18);;;/q;3*+1/p-3/t1-,2+,3+,4-,5-,6-;;;/m1.../s1

InChI key

ZVCVTWVBDIPWPZ-ZKVWPJASSA-K

Biochem/physiol Actions

Stimulates intracellular calcium mobilization.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond M Sturgeon et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(35), 7622-7634 (2018-07-25)
In the hemaphroditic sea snail, Aplysia californica, reproduction is initiated when the bag cell neurons secrete egg-laying hormone during a protracted afterdischarge. A source of depolarization for the afterdischarge is a voltage-gated, nonselective cation channel, similar to transient receptor potential
Daniela M Arduino et al.
Molecular cell, 67(4), 711-723 (2017-08-19)
The mitochondrial calcium uniporter complex is essential for calcium (Ca2+) uptake into mitochondria of all mammalian tissues, where it regulates bioenergetics, cell death, and Ca2+ signal transduction. Despite its involvement in several human diseases, we currently lack pharmacological agents for
Celeste Eno et al.
Development (Cambridge, England), 145(10) (2018-04-11)
Zebrafish germ plasm ribonucleoparticles (RNPs) become recruited to furrows of early zebrafish embryos through their association with astral microtubules ends. During the initiation of cytokinesis, microtubules are remodeled into a furrow microtubule array (FMA), which is thought to be analogous
Laurent Duchatelet et al.
Scientific reports, 10(1), 10195-10195 (2020-06-25)
The velvet belly lanternshark, Etmopterus spinax, uses counterillumination to disappear in the surrounding blue light of its marine environment. This shark displays hormonally controlled bioluminescence in which melatonin (MT) and prolactin (PRL) trigger light emission, while α-melanocyte-stimulating hormone (α-MSH) and
Hoa Thi Le et al.
Organic & biomolecular chemistry, 13(30), 8291-8297 (2015-07-04)
In order to detect small polyanions (sPAs), which play important roles in many biological systems, a triazolium cyclodextrin click cluster (5, hexakis{6-(3-methyl-4-hydroxymethyl-1H-1,2,3-triazolium-1-yl)-6-deoxy}-α-cyclodextrin iodide) was synthesized and characterized. The competition binding to 5 occupied by 5-carboxyfluorescein of inositol-1,4,5-trisphosphate (IP3), phytic acid

Questions

  1. Vorrei sapere per quanto tempo si conservano stabili le aliquote di I9766 in H20 conservate a -20°C

    1 answer
    1. Unfortunately, the solution stability has not been determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service