Skip to Content
Merck
All Photos(1)

Key Documents

I0381

Sigma-Aldrich

Isomaltotriose

≥98% (TLC)

Synonym(s):

α-D-Glucosyl-[1→6]-α-D-glucosyl-[1→6]-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (TLC)

form

lyophilized powder

technique(s)

thin layer chromatography (TLC): suitable

SMILES string

OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@H]3OC(O)[C@H](O)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16/c19-1-4-7(20)11(24)14(27)17(33-4)31-3-6-9(22)12(25)15(28)18(34-6)30-2-5-8(21)10(23)13(26)16(29)32-5/h4-29H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11+,12+,13-,14-,15-,16?,17+,18+/m1/s1

InChI key

FBJQEBRMDXPWNX-FYHZSNTMSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joana Gangoiti et al.
Biochimica et biophysica acta, 1860(6), 1224-1236 (2016-02-13)
Originally the glycoside hydrolase (GH) family 70 only comprised glucansucrases of lactic acid bacteria which synthesize α-glucan polymers from sucrose. Recently we have identified 2 novel subfamilies of GH70 enzymes represented by the Lactobacillus reuteri 121 GtfB and the Exiguobacterium
Gregory Arnal et al.
Protein science : a publication of the Protein Society, 27(6), 1093-1101 (2018-04-01)
Bacteria that reside in the mammalian intestinal tract efficiently hydrolyze dietary carbohydrates, including starch, that escape digestion in the small intestine. Starch is an abundant dietary carbohydrate comprised of α1,4 and α1,6 linked glucose, yet mammalian intestinal glucoamylases cannot effectively
Xiaolei Li et al.
International journal of biological macromolecules, 114, 235-243 (2018-03-20)
The acidophilic and thermophilic pullulanases have many potential applications in the processes of starch liquefaction and saccharification. In this study, a gene encoding an amylopullulanase from Thermofilum pendens (TPApu) was heterologously expressed in Escherichia coli. Although TPApu possessed the same
Joana Gangoiti et al.
Scientific reports, 7, 39761-39761 (2017-01-07)
Lactic acid bacteria possess a diversity of glucansucrase (GS) enzymes that belong to glycoside hydrolase family 70 (GH70) and convert sucrose into α-glucan polysaccharides with (α1 → 2)-, (α1 → 3)-, (α1 → 4)- and/or (α1 → 6)-glycosidic bonds. In recent years 3 novel subfamilies of GH70 enzymes
Karan Wangpaiboon et al.
Scientific reports, 8(1), 8340-8340 (2018-05-31)
Alternansucrase catalyses the sequential transfer of glucose residues from sucrose onto another sucrose molecule to form a long chain polymer, known as "alternan". The alternansucrase-encoding gene from Leuconostoc citreum ABK-1 (Lcalt) was successfully cloned and expressed in Escherichia coli. Lcalt

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service