Skip to Content
Merck
All Photos(1)

Documents

05389

Supelco

Isomaltose

analytical standard

Synonym(s):

6-O-α-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93355
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D 107.0±4.0°, 24 hr, c = 1 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](OC[C@H]2OC(O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(14)8(17)10(19)12(23-3)21-2-4-6(15)7(16)9(18)11(20)22-4/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11?,12+/m1/s1

InChI key

DLRVVLDZNNYCBX-RTPHMHGBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Isomaltose for research in glycan analog synthesis: Isomaltose is used in pioneering studies to develop linkage-editing pseudo-glycans, employing reductive alpha-fluorovinyl-C-glycosylation strategies. This research opens new pathways for creating glycan analogs with modified biological activities, critical in pharmaceutical research and biotechnology (Moriyama et al., 2024).

  • Isomaltose biochemical assay in food science: Rapid identification and quantification techniques for disaccharide isomers like isomaltose are developed using ESI-MS/MS. These methodologies are applied to investigate food products such as yellow rice wine, demonstrating isomaltose′s role in food chemistry and quality analysis (Yuan et al., 2023).

  • Isomaltose glycosidic bond study in carbohydrate research: Research on minor carbohydrates, including isomaltose, as authenticity markers in grape must highlights its importance in the control and quality assessment of food products, facilitating better regulatory compliance and product authentication (Paolini et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service