Key Documents
HS29025E
Microtube Rack
yellow
Select a Size
5 EACH
$79.10
$79.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
5 EACH
$79.10
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
$79.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
yellow
feature
to hold 80 tube(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS29025E
L × W × H
225 mm × 67 mm × 28 mm
Related Categories
General description
The 80-Well Microtube Rack is a popular fraction-collector-plate style rack.
Features and Benefits
  • Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
  • Frosted labeling area on bottom of rack
  • Embossed Index Numbers for quick sample ID and retrieval
  • Made of polypropylene
  • Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)