Skip to Content
Merck
All Photos(1)

Key Documents

HS23229B

Cryogenic Vial Rack

to hold, 50 vials per rack, blue

Synonym(s):

cryogenic vial rack, cryostorage rack, cryovial rack, dewar rack, liquid nitrogen rack

Sign Into View Organizational & Contract Pricing

Select a Size

4 EACH
$158.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
4 EACH
$158.00

About This Item

UNSPSC Code:
41122806
NACRES:
NB.51

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

blue
polycarbonate

feature

to hold 2 ‑ 5 mL tubes
to hold 50 vials per rack

packaging

pack of 4 ea

manufacturer/tradename

Heathrow Scientific HS23229B

L × W × H

199 mm × 105 mm × 28 mm

General description

The Cryogenic Vial rack safely stores and transports cryogenic vials.

Features and Benefits

  • Made from polycarbonate
  • Drainage holes in each well aid in cleaning
  • Alphanumeric identification for quick location and retrieval of tubes • Will withstands cryogenic freezing down to -196ºC
  • Well tabs allow one-handed opening and closing of vials that have a slot feature
  • Embossed index numbers and squared, upper left corner provide easy orientation
  • Ribbed grips make unscrewing caps one-handed, as well as handling with gloves easier
  • Autoclavable

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service