Skip to Content
Merck
All Photos(5)

Key Documents

HPA045727

Sigma-Aldrich

Anti-SDHD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PGL, Anti-PGL1, Anti-cybS, Anti-Succinate Dehydrogenase Complex, Subunit D, Integral Membrane Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RTPVVRPAHISAFLQDRPIPEWCGVQHIHLSPSHHSGSKAASLHW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDHD(6392)

Related Categories

General description

Succinate dehydrogenase complex subunit D (SDHD) gene encodes for a protein SDHD. It is a small sub unit of the cytochrome b, that is involved in the complex II of mitochondrial respiration. It is located at 11q23.1 on the human chromosome.

Immunogen

Succinate dehydrogenase complex subunit D recombinant protein epitope signature tag (PrEST)

Application

Anti-SDHD antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Mutation in this gene leads to paraganglioma and phaeochromocytoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84465

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo detection of succinate by magnetic resonance spectroscopy as a hallmark of SDHx mutations in paraganglioma
Lussey L C, et al.
Clinical Cancer Research, 22(5), 1120-1129 (2016)
Risk assessment of maternally inherited SDHD paraganglioma and phaeochromocytoma
Burnichon N, et al.
Journal of medical Genetics, 54(2), 125-133 (2017)
Analysis of the SDHD gene, the susceptibility gene for familial paraganglioma syndrome (PGL1), in pheochromocytomas
Aguiar R C T, et al.
The Journal of Clinical Endocrinology and Metabolism, 86(6), 2890-2894 (2001)
Mélanie Menara et al.
The Journal of clinical endocrinology and metabolism, 100(2), E287-E291 (2014-11-19)
Pheochromocytomas (PCC) and paragangliomas (PGL) may be caused by a germline mutation in 12 different predisposing genes. We previously reported that immunohistochemistry is a useful approach to detect patients harboring SDHx mutations. SDHA immunostaining is negative in SDHA-mutated tumors only
Charlotte Lussey-Lepoutre et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(5), 1120-1129 (2015-10-23)
Germline mutations in genes encoding mitochondrial succinate dehydrogenase (SDH) are found in patients with paragangliomas, pheochromocytomas, gastrointestinal stromal tumors, and renal cancers. SDH inactivation leads to a massive accumulation of succinate, acting as an oncometabolite and which levels, assessed on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service