Merck
  • In Vivo Detection of Succinate by Magnetic Resonance Spectroscopy as a Hallmark of SDHx Mutations in Paraganglioma.
In Vivo Detection of Succinate by Magnetic Resonance Spectroscopy as a Hallmark of SDHx Mutations in Paraganglioma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-10-23)
Charlotte Lussey-Lepoutre, Alexandre Bellucci, Aurélie Morin, Alexandre Buffet, Laurence Amar, Maxime Janin, Chris Ottolenghi, Franck Zinzindohoué, Gwennhael Autret, Nelly Burnichon, Estelle Robidel, Benjamin Banting, Sébastien Fontaine, Charles-André Cuenod, Paule Benit, Pierre Rustin, Philippe Halimi, Laure Fournier, Anne-Paule Gimenez-Roqueplo, Judith Favier, Bertrand Tavitian
ABSTRACT
Germline mutations in genes encoding mitochondrial succinate dehydrogenase (SDH) are found in patients with paragangliomas, pheochromocytomas, gastrointestinal stromal tumors, and renal cancers. SDH inactivation leads to a massive accumulation of succinate, acting as an oncometabolite and which levels, assessed on surgically resected tissue are a highly specific biomarker of SDHx-mutated tumors. The aim of this study was to address the feasibility of detecting succinate in vivo by magnetic resonance spectroscopy. A pulsed proton magnetic resonance spectroscopy ((1)H-MRS) sequence was developed, optimized, and applied to image nude mice grafted with Sdhb(-/-) or wild-type chromaffin cells. The method was then applied to patients with paraganglioma carrying (n = 5) or not (n = 4) an SDHx gene mutation. Following surgery, succinate was measured using gas chromatography/mass spectrometry, and SDH protein expression was assessed by immunohistochemistry in resected tumors. A succinate peak was observed at 2.44 ppm by (1)H-MRS in all Sdhb(-/-)-derived tumors in mice and in all paragangliomas of patients carrying an SDHx gene mutation, but neither in wild-type mouse tumors nor in patients exempt of SDHx mutation. In one patient, (1)H-MRS results led to the identification of an unsuspected SDHA gene mutation. In another case, it helped define the pathogenicity of a variant of unknown significance in the SDHB gene. Detection of succinate by (1)H-MRS is a highly specific and sensitive hallmark of SDHx mutations. This noninvasive approach is a simple and robust method allowing in vivo detection of the major biomarker of SDHx-mutated tumors.