Skip to Content
Merck
All Photos(5)

Documents

HPA039557

Sigma-Aldrich

Anti-TAZ antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BTHS, Anti-CMD3A, Anti-EFE, Anti-EFE2, Anti-G4.5, Anti-Tafazzin, Anti-XAP-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

WHVGMNDVLPNSPPYFPRFGQKITVLIGKPFSALPVLERLRAENKSAVEMRKALTDFIQEEFQHLKTQAEQLHNHLQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAZ(6901)

General description

Tafazzin (TAZ) is encoded by the WW domain containing transcription regulator 1 (WWTR1) gene mapped to human chromosome Xq28. The encoded protein contains 400 amino acids. It is characterized by a conserved WW domain, implicated in binding PPXY motif, a coiled-coil region associated with protein-protein interaction and a C-terminal motif involved in binding the PDZ domain.

Immunogen

tafazzin recombinant protein epitope signature tag (PrEST)

Application

Anti-TAZ antibody produced in rabbit has been used in immunohistochemistry and immunoprecipitation.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Tafazzin (TAZ) plays a vital role in the various cellular process including cell proliferation, differentiation, apoptosis, migration, invasion, epithelial-mesenchymal transition (EMT) and stemness in multiple human cancers. TAZ associates with other transcription factors such as RUNX2 (runt-related transcription factor 2), PPAR (peroxisome proliferator-activated receptor), PAX3 and 8 (paired box gene 3 and 8) and TTF1 (thyroid transcription factor 1) and plays an essential role in osteoblastic, myogenic and adipogenic differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77964

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aerobic glycolysis tunes YAP/TAZ transcriptional activity.
Enzo E, et.al.
The Embo Journal, 34, 1349-1370 (2015)
Yugang Wen et al.
Journal of personalized medicine, 10(4) (2020-10-18)
Radiation therapy has long been contemplated as an important mode in the treatment of rectal cancer. However, there are few ideal tools available for clinicians to make a radiotherapy decision at the time of diagnosis for rectal cancer. The purpose
Intra-individual plasticity of the TAZ gene leading to different heritable mutations in siblings with Barth syndrome.
Ferri L
European Journal of Human Genetics, 23, 1708-1712 (2015)
Regulation of TAZ in cancer.
Zhou X and Lei QY
Protein & cell, 7, 548-561 (2016)
Tafazzin protein expression is associated with tumorigenesis and radiation response in rectal cancer: a study of Swedish clinical trial on preoperative radiotherapy.
Pathak S
PLoS ONE, 9 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service