Skip to Content
Merck
All Photos(1)

Documents

HPA026829

Sigma-Aldrich

Anti-FKBP14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FK506 binding protein 14, Anti-FKBP22, Anti-FLJ20731

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ALIPEPEVKIEVLQKPFICHRKTKGGDLMLVHYEGYLEKDGSLFHSTHKHNNGQPIWFTLGILEALKGWDQGLKGMCVG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FKBP14(55033)

General description

The gene FK506 binding protein 14 (FKBP14) is mapped to human chromosome 7p15.1. It codes for a 22kDa protein expressed mainly in the endoplasmic reticulum. The encoded protein belongs to the FK506-binding family of peptidyl-prolyl cis-trans isomerases. FKBP22 is characterized with two domains, FKBP, which includes single FKBP-type fold, and an EF-hand domain with two EF-hand motifs.

Immunogen

FK506 binding protein 14, 22 kDa recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mutations in FK506 binding protein 14 (FKBP14) cause a variant of ehlers-danlos syndrome characterized with progressive kyphoscoliosis, myopathy and hearing impairment. The members of FK506-binding protein (FKBP) family facilitates various biochemical processes, such as protein folding, receptor signaling, protein trafficking and transcription. FKBP14 regulates proteins involved in notch signaling pathway. Aberration or deficiency of this gene causes specific defects in eye, bristle and wing development in drosophila.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72571

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of human peptidyl-prolyl cis-trans isomerase FKBP22 containing two EF-hand motifs.
Boudko SP
Protein Science, 23(1), 67-75 (2014)
Diana L van de Hoef et al.
Development (Cambridge, England), 140(4), 810-819 (2013-01-16)
Presenilins were identified as causative factors in familial Alzheimer's disease and also play an essential role in Notch signaling during development. We previously identified FKBP14, a member of the family of FK506-binding proteins (FKBPs), as a modifier of Presenilin in
Matthias Baumann et al.
American journal of human genetics, 90(2), 201-216 (2012-01-24)
We report on an autosomal-recessive variant of Ehlers-Danlos syndrome (EDS) characterized by severe muscle hypotonia at birth, progressive scoliosis, joint hypermobility, hyperelastic skin, myopathy, sensorineural hearing impairment, and normal pyridinoline excretion in urine. Clinically, the disorder shares many features with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service