Skip to Content
Merck

Skip To

MABC552

Anti-Kallikrein-1/KLK1 Antibody, clone 11C7.1

clone 11C7.1, from mouse

Synonym(s):

Kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Tissue kallikrein, Kallikrein-1/KLK1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
11C7.1, monoclonal
Application:
IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11C7.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KLK1(3816)

General description

Kallikrein-1 (EC:3.4.21.35; UniProt P06870; also known as Glandular kallikrein 1, hK1, Kallikrein serine protease 1, Kidney/pancreas/salivary gland kallikrein, Renal/pancreas/salivary kallikrein 1, Tissue kallikrein) is encoded by the KLK1 (also known as KLKR) gene (Gene ID 3816) in human. Kallikrein-1 is a member of the kallikrein subfamily of serine proteases, members of which are involved in diverse physiological functions and many are used clinically as biomarkers for cancer. Kallikrein-1 is initially produced as a preproprotein with a signal (aa 1-18) and a propeptide (aa 19-24), cleavage of which releases the mature protein (aa 25-262). Kallikrein-1 is the major kallikrein expressed in the kidney, where it generates kinins (e.g. lysyl-bradykinin and bradykinin) and plays role in kidney function and blood pressure regulation through vasodilatory and natriuretic effects. Decreased kidney Kallikrein-1 level is reported in experimental settings of hypertension, diabetic nephropathy, high salt- and aminoglycoside-induced kidney injury.
~30 kDa observed. 28.9 kDa/isoform 1 and 23.8 kDa/isoform 2 calculated.

Immunogen

GST-tagged recombinant protein corresponding to the C-terminal half of human Kallikrein-1/KLK1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Kallikrein-1/KLK1 in human pancreas and human colon tissue.
This Anti-Kallikrein-1/KLK1 Antibody, clone 11C7.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Kallikrein-1/KLK1.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in human salivary gland tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Kallikrein-1/KLK1 in 10 µg of human salivary gland tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC971MABN2430M
species reactivity

human

species reactivity

human

species reactivity

human

clone

11C7.1, monoclonal

clone

33.1, monoclonal

clone

83-1, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

mouse

biological source

mouse

biological source

mouse

Gene Information

human ... KLK1(3816)

Gene Information

human ... KLK13(26085)

Gene Information

human ... KLK7(5650)

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

A major focus of breast cancer research is to understand the mechanisms responsible for disease progression and drug resistance. Toward that end, it has been found that approximately two thirds of all human breast carcinomas overexpress the Estrogen Receptor α (ERα) protein and it remains the primary pharmacological target for endocrine therapy1,2. The normal cellular function of ERα is as a transcription factor that mediates a wide variety of physiological processes, many of which are dependent upon phosphorylation of the receptor at specific amino acid residues3,4. Indeed, ERα is known to be phosphorylated at a multitude of different sites, yet how these all correlate to disease remains unclear5. Here, we interrogated multiple sites of ERα for phosphorylation status by screening an extensive panel of different breast cancer patient samples and other non-breast cancer tissue microarray (TMA) slide samples to determine their relevance to disease.





Global Trade Item Number

SKUGTIN
MABC55204055977276978

Questions

Reviews

No rating value

Active Filters