Skip to Content
Merck
All Photos(2)

Key Documents

HPA023720

Sigma-Aldrich

Anti-ESRP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-RNA-binding motif protein 35A, Anti-RNA-binding protein 35A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:50- 1:200

immunogen sequence

QLHVRQILHPEASKKNVLLPECFYSFFDLRKEFKKCCPGSPDIDKLDVATMTEYLNFEKSSSVSRYGASQVEDMGNIILAMISEPYNHRFSDPERVNYKFES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ESRP1(54845)

General description

ESRP1 is a regulatory protein that mediates the splicing of endogenous FGFR2 to generate the epithelial cell isoform. Inhibition of ESRP1 results in the mesenchymal isoform of FGFR2 during splicing. ESRP1 also modulates the splicing transition from mesenchymal to epithelial isoforms for hMena, CD44 and CTNND1 . Anti-ESRP1 antibody is specific for ESRP1 in humans.

Immunogen

RNA-binding protein 35A recombinant protein epitope signature tag (PrEST)

Application

Anti-ESRP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-ESRP1 antibody is suitable for use in indirect immunofluorescence.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76397.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Jeong et al.
Oncogenesis, 6(10), e389-e389 (2017-10-11)
Epithelial splicing regulatory protein 1 (ESRP1) and 2 (ESRP2), epithelial cell-specific regulators of alternative splicing, are downregulated during the epithelial-mesenchymal transition (EMT). These factors have roles in tumor progression and metastasis in some cancers; however, their expression and function in
Morton Freytag et al.
BMC cancer, 20(1), 1220-1220 (2020-12-20)
Epithelial splicing regulatory protein 1 (ESRP1) and 2 (ESRP2) regulate alternative splicing events of various pre-mRNAs. Some of these targets play a role in cancer-associated processes, including cytoskeleton reorganization and DNA-repair processes. This study was undertaken to estimate the impact
Pietro Fici et al.
Oncotarget, 8(2), 2423-2436 (2016-12-03)
Epithelial-to-mesenchymal transition (EMT) has been shown to be associated with tumor progression and metastasis. During this process in breast cancer, a crucial role is played by alternative splicing systems. To identify a new early prognostic marker of metastasis, we evaluated
Melissa Stinnesbeck et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 129(4), 204-212 (2021-01-18)
The aim of this study was to validate prostate cancer-associated genes on transcript level and to assess the prognostic value of the most promising markers by immunohistochemistry. Based on differentially expressed genes found in a previous study, 84 genes were
Shaghayegh Saeidi et al.
PloS one, 13(1), e0190925-e0190925 (2018-01-13)
ESRP1 regulates alternative splicing, producing multiple transcripts from its target genes in epithelial tissues. It is upregulated during mesenchymal to epithelial transition associated with reprogramming of fibroblasts to iPS cells and has been linked to pluripotency. Mouse fetal germ cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service