Merck
Prognostic role of TSPAN1, KIAA1324 and ESRP1 in prostate cancer.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2021-01-18)
Melissa Stinnesbeck, Anna Kristiansen, Jörg Ellinger, Stefan Hauser, Lars Egevad, Yuri Tolkach, Glen Kristiansen
ABSTRACT
The aim of this study was to validate prostate cancer-associated genes on transcript level and to assess the prognostic value of the most promising markers by immunohistochemistry. Based on differentially expressed genes found in a previous study, 84 genes were further validated using mRNA expression data and follow-up information from the Cancer Genome Atlas (TCGA) prostate cancer cohort (n = 497). Immunohistochemistry was used for validation of three genes in an independent, clinically annotated prostatectomy patient cohort (n = 175) with biochemical relapse as endpoint. Also, associations with clinicopathological variables were evaluated. Eleven protein-coding genes from the list of 84 genes were associated with biochemical recurrence-free survival on mRNA expression level in multivariate Cox-analyses. Three of these genes (TSPAN1, ESRP1 and KIAA1324) were immunohistochemically validated using an independent cohort of prostatectomy patients. Both ESRP1 and KIAA1324 were independently associated with biochemical recurrence-free survival. TSPAN1 was univariately prognostic but failed significance on multivariate analysis, probably due to its strong correlation with high Gleason scores. Multistep filtering using the publicly available TCGA cohort, data of an earlier expression profiling study which profiled 3023 cancer-associated transcripts in 42 primary prostate cancer cases, identified two novel candidate prognostic markers (ESRP1 and KIAA1324) of primary prostate cancer for further study.