Skip to Content
Merck
All Photos(6)

Key Documents

HPA020856

Sigma-Aldrich

Anti-BCL2L12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bcl-2-like 12 protein, Anti-Bcl-2-related proline-rich protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PSESPRPCSLPIRPCYGLEPGPATPDFYALVAQRLEQLVQEQLKSPPSPELQGPPSTEKEAILRRLVALLEEEAEVINQK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2L12(83596)

General description

The gene BCL2L12 (Bcl-2-like protein 12) is mapped to human chromosome 19q13.3. It belongs to Bcl2 family of proteins.

Immunogen

Bcl-2-related proline-rich protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BCL2L12 (Bcl-2-like protein 12) binds with apoptosis regulator BCL-XL (BCL2-like 1) and BCL2 (B-cell CLL/lymphoma 2), and exhibits anti-apoptotic property. It is up-regulated in malignant bladder cancer tissue and primary glioblastoma. In acute myeloid leukemia, higher levels of BCL2L12 increase the risk of relapse.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74190

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Scorilas et al.
Genomics, 72(2), 217-221 (2001-06-13)
Members of the Bcl-2 family of apoptosis-regulating proteins contain at least one of the four evolutionarily conserved domains, termed BH1, BH2, BH3, or BH4. Here, we report the identification, cloning, physical mapping, and expression pattern of BCL2L12, a novel gene
Hellinida Thomadaki et al.
Clinical biochemistry, 45(16-17), 1362-1367 (2012-06-26)
BCL2L12 is a recently discovered and cloned gene from members of our research team. It is a novel member of the BCL2 gene family, members of which are implicated in different hematological malignancies. In the present study, we investigated and
Spyros Foutadakis et al.
Urologic oncology, 32(1), 39-39 (2013-06-25)
More than half of the diagnosed patients with bladder cancer (BCa) recur at least once following their initial treatment. Thus, patients' monitoring and prognosis is of utmost importance. However, the need for intensive surveillance of BCa significantly burdens patients' health-related
Mahdieh Sadat Taghavi et al.
In vitro cellular & developmental biology. Animal, 49(6), 465-472 (2013-05-28)
Glioblastoma progression is mainly characterized by intense apoptosis resistance and marked necrosis. Over-expression of BCL2L12, a novel member of Bcl-2 family has been shown in primary glioblastoma. BCL2L12 blocks effective caspase-3/7 maturation and inhibits p53 tumor suppressor, deriving resistance toward
Ming-Chang Yang et al.
International journal of oncology, 46(3), 1304-1316 (2015-01-15)
Bcl2L12 as a new member of the Bcl2 family, which contains a BH2 domain and shares a lower amino acid similarity with other Bcl2 family proteins. Bcl2L12 is reported to be involved in apoptosis regulation, but this role remains controversial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service