Merck
  • Bcl2L12 with a BH3-like domain in regulating apoptosis and TMZ-induced autophagy: a prospective combination of ABT-737 and TMZ for treating glioma.
Bcl2L12 with a BH3-like domain in regulating apoptosis and TMZ-induced autophagy: a prospective combination of ABT-737 and TMZ for treating glioma.
International journal of oncology (2015-01-15)
Ming-Chang Yang, Joon-Khim Loh, Yi-Yang Li, Wen-Sheng Huang, Chia-Hua Chou, Jiin-Tsuey Cheng, Yeng-Tseng Wang, Ann-Shung Lieu, Shen-Long Howng, Yi-Ren Hong, An-Kuo Chou
ABSTRACT
Bcl2L12 as a new member of the Bcl2 family, which contains a BH2 domain and shares a lower amino acid similarity with other Bcl2 family proteins. Bcl2L12 is reported to be involved in apoptosis regulation, but this role remains controversial in different cancer type. Temozolomide (TMZ) is currently used to intervene glioma multiforme (GBM), but an acquired chemotherapeutic resistance maybe occurred due to undesired autophagy. Previous studies uncovered that Bcl2L12 may interact with Bcl-xL and may harbor a BH3-like domain. Therefore, we investigated whether this BH3-like domain is responsible for the Bcl2L12 anti-apoptotic property. Moreover, we tested whether ABT-737, a BH3 mimetic agent, can be combined with TMZ to treat GBM. We aligned Bcl2L12 with Bcl2 family members, compared interacting pattern of BH3 domain and their protein 3D structure. We identified that Bcl2L12 interacts with Bcl-xL and Bcl2 in yeast two-hybrid system. Bcl2L12192-220 was a minimal region for Bcl2L12-Bcl-xL interaction. Five-point mutations with respect to hydrophobic and charge residues were generated to test whether they are the key residue of BH3-like domain. Our data showed that both h1 (L213) and h2 residue (L217) are essential for Bcl2L12 interacting with Bcl2 family proteins. Ectopically expressed h1 or h2 mutant in U87MG cell line resulted in reactivation of cleaved-PARP, caspase-3 and cytochrome c releasing compared to Bcl2L12 wt group. Implementing ABT-737 combined with TMZ provided a superior effect on apoptosis induction in Bcl2L12 wt group, which effectively reactivated apoptotic markers. Altogether, our findings indicated that Bcl2L12 retains a BH3-like domain, which is important for the Bcl2L12 anti-apoptotic property and TMZ-induced autophagy. Our results basically support the idea of using ABT-737 to counteract the anti-apoptotic role of Bcl2L12 and sensitize drug response of the GBM cells to TMZ.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%