Skip to Content
Merck
All Photos(3)

Key Documents

HPA017343

Sigma-Aldrich

Anti-PALD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1274, Anti-Paladin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GEFQVVMKVVQLLPDGHRVKKEVDAALDTVSETMTPMHYHLREIIICTYRQAKAAKEAQEMRRLQLRSLQYLERYVCLILFNAYLHLEKADSWQRPFSTWMQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Phosphatase domain containing, paladin 1 (PALD1) is a putative cytoplasmic microfilament-associated phosphoprotein consisting of four predicted protein phosphatase (PTP) domains. It is expressed in the vasculature, hematopoietic cells and other nonvascular cells.

Immunogen

Paladin recombinant protein epitope signature tag (PrEST)

Application

Anti-KIAA1274 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Phosphatase domain containing, paladin 1 (PALD1) inhibits insulin′s ability to downregulate a FOXO1A-driven reporter gene, suppresses upstream insulin-stimulated AKT (serine/threonine kinase) phosphorylation and reduces insulin receptor (IR) abundance when overexpressed. Whereas knockdown of this gene causes the reverse. It has been suggested that PALD1 may act as both positive and negative control during maintenance of proper skeletal myogenic differentiation in vitro.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72575

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Min A Huang et al.
PloS one, 4(9), e6871-e6871 (2009-09-04)
Type 2 diabetes develops due to a combination of insulin resistance and beta-cell failure and current therapeutics aim at both of these underlying causes. Several negative regulators of insulin signaling are known and are the subject of drug discovery efforts.
Elena A May et al.
The Journal of cell biology, 220(5) (2021-04-16)
The primary cilium is a signaling compartment that interprets Hedgehog signals through changes of its protein, lipid, and second messenger compositions. Here, we combine proximity labeling of cilia with quantitative mass spectrometry to unbiasedly profile the time-dependent alterations of the
Elisabet Wallgard et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 241(4), 770-786 (2012-02-23)
Angiogenesis is implicated in many pathological conditions. The role of the proteins involved remains largely unknown, and few vascular-specific drug targets have been discovered. Previously, in a screen for angiogenesis regulators, we identified Paladin (mouse: X99384, human: KIAA1274), a protein
Ngoc-Uyen-Nhi Nguyen et al.
PloS one, 10(4), e0124762-e0124762 (2015-04-16)
Palladin is a microfilament-associated phosphoprotein whose function in skeletal muscle has rarely been studied. Therefore, we investigate whether myogenesis is influenced by the depletion of palladin expression known to interfere with the actin cytoskeleton dynamic required for skeletal muscle differentiation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service