Skip to Content
Merck
All Photos(4)

Key Documents

HPA014572

Sigma-Aldrich

Anti-BMP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BMP-1, Anti-Bone Morphogenetic Protein 1 precursor, Anti-Mammalian tolloid protein, Anti-PCP, Anti-Procollagen C-proteinase, Anti-mTld

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

DSEPLNYKDPCKAAAFLGDIALDEEDLRAFQVQQAVDLRRHTARKSSIKAAVPGNTSTPSCQSTNGQPQRGACGRWRGRSRSRRAATSRPERVWPDGVIPFVIGGNFTGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP1(649)

General description

BMP1 (bone morphogenetic protein 1) is a metalloproteinase belonging to the astacin family. This gene is localized to human chromosome 8p21.3, and is alternatively spliced to produce BMP1 and the longer isoform tolloid protein (TLD). This protein is produced as a proenzyme, which consists of leader peptide, a prodomain, one catalytic domain, three CUB domains, and one epidermal growth factor (EGF)-like domain.

Immunogen

Bone morphogenetic protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BMP1 (bone morphogenetic protein 1) processes the naïve form of extracellular matrix (ECM) proteins to the mature form, and therefore, are involved in morphogenesis. It also controls the processing of multiple growth factors such as, TGFβ, BMP2, BMP4 and GFD (growth differentiation factor) 8. It facilitates the conversion of bone marrow mesenchymal stem cells to mineralized bone cells. Thus, it enhances bone repair. It makes up the BMP1/TLD-like (tolloid-like) complex, which also includes TLD, tolloid-like protein 1 and tolloid-like protein 2. It is involved in the processing of procollagen I C-terminal propeptide (PICP), type II, type III, and type V procollagen and lysyl oxidase zymogen. Mutation in this gene is linked to autosomal recessive osteogenesis imperfecta. During embryogenesis, it facilitates the binding of BMPs to cognate receptors by degrading the BMP-antagonist chordin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72700

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana Kovacevic Vojtusek et al.
Diagnostics (Basel, Switzerland), 12(3) (2022-03-26)
We have previously shown that metzincin protease ADAMTS-4 accompanies renal fibrogenesis, as it appears in the blood of hemodialysis patients. Native kidney (NKB) and kidney transplant (TXCI) biopsy samples as well as plasma from patients with various stages of CKD
Víctor Martínez-Glez et al.
Human mutation, 33(2), 343-350 (2011-11-05)
Herein, we have studied a consanguineous Egyptian family with two children diagnosed with severe autosomal recessive osteogenesis imperfecta (AR-OI) and a large umbilical hernia. Homozygosity mapping in this family showed lack of linkage to any of the previously known AR-OI
Mat Leighton et al.
The Journal of biological chemistry, 278(20), 18478-18484 (2003-03-15)
Bone morphogenetic protein (BMP)-1 is a zinc-dependent metalloproteinase that cleaves a variety of extracellular matrix substrates, including type I procollagen. Little is known about the site of action of BMP-1, although the extracellular matrix seems likely to be it. BMP-1
Lovorka Grgurevic et al.
Biochemical and biophysical research communications, 408(1), 25-31 (2011-04-02)
Members of the astacin family of metalloproteinases such as human bone morphogenetic protein 1 (BMP-1) regulate morphogenesis by processing precursors to mature functional extracellular matrix (ECM) proteins and several growth factors including TGFβ, BMP2, BMP4 and GFD8. We have recently
Yung-Yu Hsieh et al.
BMC cancer, 18(1), 508-508 (2018-05-04)
Gastric cancer is the eighth most common cancer in Taiwan, with a 40% 5-year survival rate. Approximately 40% of patients are refractory to chemotherapy. Currently, the anti-HER2 therapy is the only clinically employed targeted therapy. However, only 7% patients in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service