Skip to Content
Merck
All Photos(6)

Key Documents

HPA013972

Sigma-Aldrich

Anti-RHBDD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Rhomboid domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PLKKIMEACAGGFSSSVGYPGRQYYFNSSGSSGYQDYYPHGRPDHYEEAPRNYDTYTAGLSEEEQLERALQASLWDRGNTRNSPPPYGFHLSPEEMRRQRLHRFD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHBDD1(84236)

General description

The gene RHBDD1 (rhomboid domain containing 1) is a serine protease belonging to the Rhomboid family of proteins. The protein contains a rhomboid domain and is highly expressed in the testis. The gene is mapped to human chromosome 2q36.3.

Immunogen

Rhomboid domain-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-RHBDD1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The protein rhomboid domain containing 1 cleaves the Bcl-2 protein Bik, a proapoptotic member, and regulates Bik-mediated apoptotic pathway. It plays a crucial role in the progression of several cancers and may serve as a therapeutic target. It induces proteolysis of TSAP6 (tumor suppressor activated pathway-6), a multi-pass transmembrane protein, and regulates exosomal trafficking.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72901

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Chen et al.
Folia neuropathologica, 60(2), 195-209 (2022-08-12)
This study was designed to elucidate the relationship of miR-211-3p and rhomboid domain containing 1 (RHBDD1) in glioma. Here, we first observed that miR-211-3p directly targets the 3˘-UTR of RHBDD1 in glioma cells using dual-luciferase reporter assay, RNA immunoprecipitation (RIP)
Junyi Han et al.
Molecular medicine reports, 12(1), 377-381 (2015-02-20)
Rhomboid domain containing 1 (RHBDD1), is a member of the rhomboid protease family, which has a pivotal role in the progression of numerous severe malignancies. However, its role in colorectal carcinoma (CRC) remains to be elucidated. In the present study
Akira Meguro et al.
PloS one, 7(6), e39300-e39300 (2012-06-23)
Lattice degeneration of the retina is a vitreoretinal disorder characterized by a visible fundus lesion predisposing the patient to retinal tears and detachment. The etiology of this degeneration is still uncertain, but it is likely that both genetic and environmental
Kyojiro N Ikeda et al.
The Journal of biological chemistry, 294(30), 11486-11497 (2019-06-10)
Rhomboid-like proteins are evolutionarily conserved, ubiquitous polytopic membrane proteins, including the canonical rhomboid intramembrane serine proteases and also others that have lost protease activity during evolution. We still have much to learn about their cellular roles, and evidence suggests that
Chunhua Wan et al.
PloS one, 7(5), e37452-e37452 (2012-05-25)
We have previously reported that rhomboid domain containing 1 (RHBDD1), a mammalian rhomboid protease highly expressed in the testis, can cleave the Bcl-2 protein Bik. In this study, we identified a multi-pass transmembrane protein, tumor suppressor activated pathway-6 (TSAP6) as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service