Skip to Content
Merck
All Photos(2)

Key Documents

HPA013870

Sigma-Aldrich

Anti-CMTM3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CKLF-like MARVEL transmembrane domain-containing protein 3, Anti-Chemokine- like factor superfamily member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

NDVAKFLKQGDSADETTAHKTEEENSDSDS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CMTM3(123920)

General description

The gene CMTM3 (CKLF-like MARVEL transmembrane domain containing 3) encodes a member of the CMTM family that resembles chemokines and the transmembrane-4 superfamily of proteins. It is broadly expressed in normal human adult tissues but is highly expressed in the testes. Its expression is found to be reduced in carcinoma cell lines and primary tumors. The gene is localized to human chromosome 16q21. The encoded protein contains one leucine zipper and two LXXLL motifs.

Immunogen

CKLF-like MARVEL transmembrane domain-containing protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein CKLF-like MARVEL transmembrane domain containing 3 functions as a tumor suppressor. Its expression in tumor cells leads to caspase-3 activation, resulting in suppression of cell growth and apoptosis. Its overexpression is found to inhibit AR (androgen receptor) transcription activity. It may function in the maturation and maintenance of the male reproductive system. It is also highly expressed in peripheral blood mononuclear cells (PBMCs), suggesting its role in immune system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72664

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zhang et al.
Gut pathogens, 15(1), 6-6 (2023-02-15)
CKLF-like MARVEL transmembrane domain containing 3 (CMTM3) plays an important role in cancer development. Although Helicobacter pylori (H. pylori) infection is a main cause of gastric cancer, the function of CMTM3 during H. pylori infection remains unclear. CMTM3 expression levels
Yu Wang et al.
Biochemical and biophysical research communications, 371(1), 54-58 (2008-04-12)
CMTM is a novel family of proteins linking chemokines and TM4SF. Several members of this family are highly expressed in testes and regulate androgen receptor (AR) transcription activity. One member of this family, CMTM3, has the highest expression level in
Ji Zhong et al.
Journal of biochemistry and molecular biology, 39(5), 537-545 (2006-09-28)
CMTM/CKLFSF is a novel family of proteins linking chemokines and TM4SF. In humans, these proteins are encoded by nine genes, CKLF and CMTM1-8/CKLFSF1-8. Here we report the characteristics and expression profile of CMTM3/CKLFSF3. Human CMTM3/CKLFSF3 has a high sequence identity
Yu Wang et al.
Cancer research, 69(12), 5194-5201 (2009-06-11)
Closely located at the tumor suppressor locus 16q22.1, CKLF-like MARVEL transmembrane domain-containing member 3 and 4 (CMTM3 and CMTM4) encode two CMTM family proteins, which link chemokines and the transmembrane-4 superfamily. In contrast to the broad expression of both CMTM3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service