Merck
  • CMTM3 protects the gastric epithelial cells from apoptosis and promotes IL-8 by stabilizing NEMO during Helicobacter pylori infection.
CMTM3 protects the gastric epithelial cells from apoptosis and promotes IL-8 by stabilizing NEMO during Helicobacter pylori infection.
Gut pathogens (2023-02-15)
Jing Zhang, Jing Ning, Weiwei Fu, Yanyan Shi, Jing Zhang, Shigang Ding
ABSTRACT
CKLF-like MARVEL transmembrane domain containing 3 (CMTM3) plays an important role in cancer development. Although Helicobacter pylori (H. pylori) infection is a main cause of gastric cancer, the function of CMTM3 during H. pylori infection remains unclear. CMTM3 expression levels in tissues from H. pylori-infected patients and cells co-cultured with H. pylori were analyzed. qRT-PCR and ELISA were used to investigate the effects of CMTM3 on interleukin 8 (IL-8) expression. Annexin V/propidium iodide staining was performed to evaluate the function of CMTM3 in the apoptosis of gastric epithelial cells. Proteomic analysis was performed to explore the underlying mechanism of CMTM3 during H. pylori infection. The interaction between CMTM3 and NEMO was determined via co-immunoprecipitation, HA-ubiquitin pull-down assay, and immunofluorescence. H. pylori induced a significant increase in CMTM3 expression. CMTM3 inhibited gastric mucosal epithelial cells from apoptosis and increased the expression level of IL-8 during H. pylori infection. KEGG pathway enrichment analysis revealed that differentially expressed proteins were involved in epithelial cell signaling in H. pylori infection. CMTM3 directly interacted with NEMO, which promoted protein stabilization by down-regulation of its ubiquitylation. CMTM3 reduces apoptosis and promotes IL-8 expression in the gastric epithelial cells by stabilizing NEMO during H. pylori infection. These findings characterize a new role for CMTM3 in host-pathogen interactions and provide novel insight into the molecular regulation of NEMO.