Skip to Content
Merck
All Photos(7)

Key Documents

HPA004829

Sigma-Aldrich

Anti-NNT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NAD(P) transhydrogenase, mitochondrial precursor antibody produced in rabbit, Anti-Nicotinamide nucleotide transhydrogenase antibody produced in rabbit, Anti-Pyridine nucleotide transhydrogenase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ANLLKTVVTGCSCPLLSNLGSCKGLRVKKDFLRTFYTHQELWCKAPVKPGIPYKQLTVGVPKEIFQNEKRVALSPAGVQNLVKQGFNVVVESGAGEASKFSDDHYRVAGAQIQGAKEVLASDLVVKVRAPMVNPTLGVHEADLLKTSGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NNT(23530)

General description

NNT (nicotinamide nucleotide transhydrogenase) is an integral protein of the inner mitochondrial membrane. It is composed of three subunits termed dI, dII and dIII. The dI component binds to the NAD(+)/NADH, and the dIII binds to the NADP(+)/NADPH, protrude from the membrane.

Immunogen

NAD(P) transhydrogenase, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NNT (nicotinamide nucleotide transhydrogenase) acts as an antioxidant defense gene. It catalyzes the transhydrogenation reaction of transferring hydride ion from NAD(H) to NADP(H). In reverse transhydrogenation, it acts by the outward proton translocation and formation of a proton motive force. It has been reported that NNT may also participate in reactive oxygen species (ROS) detoxification in human adrenal glands. Mutations in NNT gene cause familial glucocorticoid deficiency.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86820

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack L Williams et al.
Life science alliance, 3(4) (2020-03-28)
The C57BL/6J and C57BL/6N mice have well-documented phenotypic and genotypic differences, including the infamous nicotinamide nucleotide transhydrogenase (Nnt) null mutation in the C57BL/6J substrain, which has been linked to cardiovascular traits in mice and cardiomyopathy in humans. To assess whether
Y Hatefi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 10(4), 444-452 (1996-03-01)
The energy-transducing nicotinamide nucleotide transhydrogenases of mammalian mitochondria and bacteria are structurally related membrane-bound enzymes that catalyze the direct transfer of a hydride ion between NAD(H) and NADP(H) in a reaction that is coupled to transmembrane proton translocation. The protonmotive
B Zieger et al.
DNA sequence : the journal of DNA sequencing and mapping, 7(6), 369-373 (1997-01-01)
Pyridine nucleotide transhydrogenases (E.C.1.6.1.1) are integral proteins of the inner mitochondrial membrane. These enzymes are part of the energy-transfer system of the respiratory chain and specifically catalyze the transfer of a hydride ion between nicotinamide adenine dinucleotide, NAD(H), and oxidized
S A White et al.
Structure (London, England : 1993), 8(1), 1-12 (2000-02-16)
Transhydrogenase, located in the inner membranes of animal mitochondria and the cytoplasmic membranes of bacteria, couples the transfer of reducing equivalents between NAD(H) and NADP(H) to proton pumping. The protein comprises three subunits termed dI, dII and dIII. The dII
Jozsef Gal et al.
Journal of neuropathology and experimental neurology, 77(1), 21-39 (2017-12-01)
Misfolded protein in the amygdala is a neuropathologic feature of Alzheimer disease and many other neurodegenerative disorders. We examined extracts from human amygdala (snap-frozen at autopsy) to investigate whether novel and as yet uncharacterized misfolded proteins would be detectable. Polypeptides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service