Skip to Content
Merck
All Photos(3)

Key Documents

HPA001623

Sigma-Aldrich

Anti-EFNB3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EPH-related receptor transmembrane ligand ELK-L3, Anti-EPH-related receptor tyrosine kinase ligand 8, Anti-Ephrin-B3 precursor, Anti-LERK-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HSSPNYEFYKLYLVGGAQGRRCEAPPAPNLLLTCDRPDLDLRFTIKFQEYSPNLWGHEFRSHHDYYIIATSDGTREGLESLQGGVCLTRGMKVLLRVGQSPRGGAVPRKPVSEMPMERDRGAAHSLEPGKENLPGDPTSNATS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFNB3(1949)

Immunogen

Ephrin-B3 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ephrin-B3 is a protein encoded by the EFNB3 gene in humans and is a member of the ephrin gene family. It is important in brain development as well as in its maintenance and may have profound effects on cell morphology. It binds specifically to B lymphocytes in blood. It binds by a non-classical receptor, and induces migration of the memory B cell subpopulation. It binds to cells that induce cellular signaling and influences cell rounding and cell spreading. It is found that its expression and signaling through Rac1 are critically important for glioma invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77450

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Ståhl et al.
Journal of proteome research, 10(5), 2566-2578 (2011-03-19)
The ephrin and Eph signaling circuit has been reported as deregulated in a number of tumor types including nonsmall cell lung cancer (NSCLC). Here we show that suppression of the ephrin-familly member ephrin B3 decreases NSCLC cell proliferation and has
Halvor L Holen et al.
The Biochemical journal, 433(1), 215-223 (2010-10-12)
The ephrins are a family of proteins known to bind the Eph (erythropoietin-producing hepatocellular) receptor tyrosine kinase family. In the present paper, we provide data showing that ephrin-B3 binds a sulfated cell-surface protein on HEK-293T (human embryonic kidney-293 cells expressing
H L Holen et al.
Scandinavian journal of immunology, 74(2), 144-154 (2011-03-31)
Eph receptors and ephrin ligands have been shown to be differentially expressed on leucocytes. Here, we show that one member of the ephrin-B subfamily of ephrins, ephrin-B3, specifically binds to B lymphocytes in blood. No binding was observed to T
Mitsutoshi Nakada et al.
Cancer research, 66(17), 8492-8500 (2006-09-05)
Eph receptor tyrosine kinases are involved in nervous system development. Eph ligands, termed ephrins, are transmembrane proteins that bind to Eph receptors, the mutual activation of which causes repulsive effects in reciprocally contacting cells. Previously, we showed that overexpression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service