Skip to Content
Merck
All Photos(1)

Key Documents

H5000

Sigma-Aldrich

Hexokinase from Saccharomyces cerevisiae

Type III, lyophilized powder, ≥25 units/mg protein (biuret)

Synonym(s):

ATP:D-Hexose-6-phosphotransferase, Hexokinase from yeast

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
Pricing and availability is not currently available.

type

Type III

form

lyophilized powder

specific activity

≥25 units/mg protein (biuret)

mol wt

dimer 110 kDa

UniProt accession no.

foreign activity

glucose-6-phosphate dehydrogenase and phosphoglucose isomerase ≤10%

storage temp.

−20°C

Gene Information

bakers yeast ... HXK1(850614) , HXK2(852639)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing ATP as a phosphate source.

The rate of phosphorylation varies with different hexoses (pH 7.5, 30 °C).
D-fructose KM: 0.33 mM
D-glucose KM: 0.12 mM
D-mannose KM: 0.05 mM

Yeast hexokinase exists as two similar isoforms, PI and PII (A and B), with isoelectric points of 5.25 and 4, respectively.

Molecular Weight: ~ 54 kDa (monomer)
~110 kDa (dimer)
Optimal pH: 7.5 to 9.0
Extinction Coefficient: E1% = 8.85 (PI) and 9.47 (PII) at 280 nm

Activators: Hexokinase requires Mg2+ ions (KM = 2.6 mM) for activity. Hexokinase is activated by catecholamines and related compounds.

Inhibitors: sorbose-1-phosphate, polyphosphates, 6-deoxy-6-fluoroglucose, 2-C-hydroxy-methylglucose, xylose, lyxose, and thiol reactive compounds (Hg2+ and 4-chloromercuribenzoate)

Unit Definition

One unit will phosphorylate 1.0 μmole of D-glucose per min at pH 7.6 at 25 °C, unless otherwise indicated below.

Physical form

Lyophilized powder containing approx. 15% sodium citrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seok-Hyung Kim et al.
PLoS genetics, 9(6), e1003563-e1003563 (2013-06-21)
Multiple Acyl-CoA Dehydrogenase Deficiency (MADD) is a severe mitochondrial disorder featuring multi-organ dysfunction. Mutations in either the ETFA, ETFB, and ETFDH genes can cause MADD but very little is known about disease specific mechanisms due to a paucity of animal
Charles Betz et al.
The Journal of cell biology, 203(4), 563-574 (2014-01-05)
Target of rapamycin (TOR) forms two conserved, structurally distinct kinase complexes termed TOR complex 1 (TORC1) and TORC2. Each complex phosphorylates a different set of substrates to regulate cell growth. In mammals, mTOR is stimulated by nutrients and growth factors
Markus Weiler et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 409-414 (2013-12-25)
A hypoxic microenvironment induces resistance to alkylating agents by activating targets in the mammalian target of rapamycin (mTOR) pathway. The molecular mechanisms involved in this mTOR-mediated hypoxia-induced chemoresistance, however, are unclear. Here we identify the mTOR target N-myc downstream regulated
Daniel Romaker et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(17), 6335-6340 (2014-04-16)
MicroRNAs (miRNAs) are major posttranscriptional regulators of a wide variety of biological processes. However, redundancy among most miRNAs has made it difficult to identify their in vivo functions. We previously demonstrated that global inhibition of miRNA biogenesis in Xenopus resulted
François Vaillant et al.
Cancer cell, 24(1), 120-129 (2013-07-13)
The prosurvival protein BCL-2 is frequently overexpressed in estrogen receptor (ER)-positive breast cancer. We have generated ER-positive primary breast tumor xenografts that recapitulate the primary tumors and demonstrate that the BH3 mimetic ABT-737 markedly improves tumor response to the antiestrogen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service