Skip to Content
Merck
All Photos(1)

Key Documents

G8404

Sigma-Aldrich

Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides

recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)

Synonym(s):

Entner-Doudoroff enzyme, G6PD, G6PDH, NADP glucose 6-phosphate dehydrogenase, G-6-P-DH

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$87.10
1000 UNITS
$191.00
2000 UNITS
$310.00
10000 UNITS
$1,050.00

$87.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 UNITS
$87.10
1000 UNITS
$191.00
2000 UNITS
$310.00
10000 UNITS
$1,050.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$87.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

bacterial (Leuconostoc mesenteroides)

Quality Level

recombinant

expressed in E. coli

form

ammonium sulfate suspension

specific activity

≥550 units/mg protein (biuret)

mol wt

128 kDa

storage condition

(Tightly closed)

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

foreign activity

creatine phosphokinase, glutathione reductase, myokinase, NADH oxidase, NADPH oxidase, phosphoglucomutase, 6-phosphogluconic dehydrogenase, phosphoglucose isomerase, lactic dehydrogenase, hexokinase ≤0.01%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Glucose 6-phosphate dehydrogenase (G-6-P-DH) is a key regulatory enzyme in the first step of the pentose phosphate pathway.[1] G-6-P-DH is a glycoprotein with a molecular mass of 128 kDa (gel filtration).[2]G6PD is a dimer and consists of a single active site at each subunit.[3]

Research area: Cell Signaling

Application

Glucose-6-phosphate dehydrogenase has been used:

  • to test ketose reductase activity in developing maize endosperm.[4]
  • to determine the levels of mannose in coronary heart disease patient-derived serum[5]
  • to study its activity on extracellular polymeric substance (EPS) extract to determine cell lysis through sonication[6]
  • to determine the glucose uptake in cultured human muscle satellite cells[7]

Biochem/physiol Actions

G6PD provides all cells with reducing power as nicotinamide adenine dinucleotide phosphate (NADPH) which enables the cells to balance oxidative stress. Mutations in the G6PD gene are associated with X-linked G6PD deficiency, a hereditary genetic defect such as neonatal jaundice and acute hemolytic anemia.[8]
Glucose-6-phosphate dehydrogenase (G6PD) catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose 6-phosphate to 6-phospho-D-gluconate per min in the presence of NAD at pH 7.8 at 30 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 containing 50 mM Tris and 1 mM MgCl2, pH 7.5

Other Notes

The volume is lot specific and can be calculated using the data:
For example, lot 0000114274
Units per mg Protein = 846
Mg Protein per mL - 9
Units per mL = 7614
G8404-1000U = approx 132 ul

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemang M Parikh et al.
BMC endocrine disorders, 21(1), 32-32 (2021-03-01)
Insulin resistance (IR) in skeletal muscle is a key feature of the pre-diabetic state, hypertension, dyslipidemia, cardiovascular diseases and also predicts type 2 diabetes. However, the underlying molecular mechanisms are still poorly understood. To explore these mechanisms, we related global
Angelo Minucci et al.
IUBMB life, 61(1), 27-34 (2008-10-23)
Glucose 6-phosphate dehydrogenase (G6PD) deficiency is the most common defect of red blood cells. Although some different laboratory techniques or methods are employed for the biochemical screening, a strict relationship between biochemists, clinicians, and molecular biologists is necessary for a
Charles-David Dubé et al.
AMB Express, 9(1), 23-23 (2019-02-08)
Extracellular polymeric substances (EPS) play major roles in the efficacy of biofilms such as anaerobic granules, ranging from structural stability to more specific functions. The EPS of three granular anaerobic sludges of different origins were studied and compared. Particularly, the
M D Cappellini et al.
Lancet (London, England), 371(9606), 64-74 (2008-01-08)
Glucose-6-phosphate dehydrogenase (G6PD) deficiency is the most common human enzyme defect, being present in more than 400 million people worldwide. The global distribution of this disorder is remarkably similar to that of malaria, lending support to the so-called malaria protection
Cara A Griffiths et al.
Nature, 540(7634), 574-578 (2016-12-16)
The pressing global issue of food insecurity due to population growth, diminishing land and variable climate can only be addressed in agriculture by improving both maximum crop yield potential and resilience. Genetic modification is one potential solution, but has yet

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Protocols

To measure glucose-6-phosphate dehydrogenase activity, beta-nicotinamide adenine dinucleotide phosphate is used in a spectrophotometric rate determination assay at 340 nm.

Questions

  1. Hallo, wieviel beträgt das Volumen für 1 kU? Vielen Dank.

    1 answer
    1. The volume is lot specific and can be calculated using the data provided in the Certificate of Analysis.
      For example, lot 0000114274
      Units per mg Protein = 846
      Mg Protein per mL - 9
      Units per mL = 7614
      G8404-1000U = approx 132 ul

      Please see the link below to review this Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/G8404-10KU0000241288.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service