![Hydrocortisone British Pharmacopoeia (BP) Assay Standard](/deepweb/assets/sigmaaldrich/product/structures/314/613/9abbaad6-0ed9-4b3c-9141-16d4d9f98809/640/9abbaad6-0ed9-4b3c-9141-16d4d9f98809.png)
Hydrocortisone
![Hydrocortisone British Pharmacopoeia (BP) Assay Standard](/deepweb/assets/sigmaaldrich/product/structures/314/613/9abbaad6-0ed9-4b3c-9141-16d4d9f98809/640/9abbaad6-0ed9-4b3c-9141-16d4d9f98809.png)
Synonym(s):
11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M
Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing