Skip to Content
Merck
All Photos(1)

Key Documents

H0255

Sigma-Aldrich

Hexadecane

≥99%

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
Beilstein:
1736592
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

7.8 (vs air)

vapor pressure

1 mmHg ( 105.3 °C)

Assay

≥99%

autoignition temp.

395 °F

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gibum Kwon et al.
Advanced materials (Deerfield Beach, Fla.), 24(27), 3666-3671 (2012-06-13)
In this work, the first-ever membrane-based single unit operation that enables gravity driven, on-demand separation of various oil-water mixtures is developed. Using this methodology, the on-demand separation of free oil and water, oil-in-water emulsions, and water-in-oil emulsions is demonstrated, with
Sonja Herman et al.
Autoimmunity, 45(8), 602-611 (2012-08-25)
Hydrocarbon oils such as pristane or hexadecane induce arthritis and lupus in rodents sharing clinical and pathological features with the human diseases rheumatoid arthritis and systemic lupus erythematosus, respectively. In pristane-induced lupus in the mouse induction of apoptosis and augmentation
Kata Farkas et al.
Food and environmental virology, 7(3), 261-268 (2014-10-25)
Adsorption to aquifer media is an important process in the removal of viruses from groundwater. Even though hydrophobic interactions have been shown to contribute to adsorption, little is known about the hydrophobicity of viruses found in groundwater. In this study
Jaejoon Jung et al.
Microbial ecology, 70(4), 912-921 (2015-05-10)
Red clay was previously used to enhance bioremediation of diesel-contaminated soil. It was speculated that the enhanced degradation of diesel was due to increased bacterial growth. In this study, we selected Acinetobacter oleivorans DR1, a soil-borne degrader of diesel and
Diana Gutiérrez et al.
Applied and environmental microbiology, 81(10), 3336-3348 (2015-03-10)
Phage therapy is a promising option for fighting against staphylococcal infections. Two lytic phages, vB_SauM_phiIPLA-RODI (phiIPLA-RODI) and vB_SepM_phiIPLA-C1C (phiIPLA-C1C), belonging to the Myoviridae family and exhibiting wide host ranges, were characterized in this study. The complete genome sequences comprised 142,348

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service