Skip to Content
Merck
All Photos(1)

Key Documents

128503

Sigma-Aldrich

Heptadecane

Heptadecane

99%

Synonym(s):

n-Heptadecane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$47.30
100 G
$124.00

$47.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$47.30
100 G
$124.00

About This Item

Linear Formula:
CH3(CH2)15CH3
CAS Number:
Molecular Weight:
240.47
Beilstein:
1738898
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32150101
PubChem Substance ID:
NACRES:
NA.22

$47.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

8.3 (vs air)

vapor pressure

1 mmHg ( 115 °C)

Assay

99%

refractive index

n20/D 1.436 (lit.)

bp

302 °C (lit.)

mp

20-22 °C (lit.)

density

0.777 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCC

InChI

1S/C17H36/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3-17H2,1-2H3

InChI key

NDJKXXJCMXVBJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heptadecane was used as an internal standard in the reaction to study the first direct comparison of kava lactone extraction efficiencies[1]. It was also used as the appropriate suspended solvent for the extraction and concentration of EO (essential oil)[2].

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kubátová et al.
Journal of chromatography. A, 923(1-2), 187-194 (2001-08-21)
Subcritical water extraction of lactones from a kava (Piper metlhysticum) root was compared to a Soxhlet extraction with water, to boiling in water, and to a sonication in acetone. For ground kava (250-500 microm), 2 h of subcritical water extraction
Jiao Jiao et al.
Analytica chimica acta, 804, 143-150 (2013-11-26)
A rapid, green and effective miniaturized sample preparation and analytical technique, i.e. ionic liquids-assisted microwave distillation coupled with headspace single-drop microextraction (ILAMD-HS-SDME) followed by gas chromatography-mass spectrometry (GC-MS) was developed for the analysis of essential oil (EO) in Fructus forsythiae.
Humberto Reyes-Prado et al.
Scientific reports, 10(1), 5633-5633 (2020-03-30)
Herbivory insects can discriminate the quality of a host plant for food or oviposition, by detecting the volatile organic compounds (VOC's) released by the plant, however, damaged plants may release a different VOC's profile modifying the insects' response. We tested
P Bonin et al.
Chemosphere, 41(9), 1321-1326 (2000-11-01)
The influence of different states of oxygen supply on heptadecane mineralization has been investigated in resting cell suspensions of Pseudomonas nautica. The rate of heptadecane biodegraded was constant for oxygen concentrations between 21% and 10% (v/v) (about 100% and 50%
Stephen M Long et al.
Journal of computational chemistry, 32(8), 1541-1549 (2011-02-02)
A new population-based incremental learning algorithm for conformational searching of molecules is presented. This algorithm is particularly effective at determining, by relatively small number of energy minimizations, global energy minima of large flexible molecules. The algorithm is also able to

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Viet Nam
    • Review 1
    • Votes 0
    4 out of 5 stars.

    Saleman

    well very good for your product, I hope your product with cheaper than in near future

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service