Skip to Content
Merck
All Photos(1)

Key Documents

GERPN2631

Matrix Metalloproteinase-2 (MMP-2) Biotrak Activity Assay System

Cytiva RPN2631, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN2631

shipped in

dry ice

storage temp.

−20°C

General description

Biotrak MMP-2, Activity Assay System provides a simple, specific and precise quantitative determination of active or pro MMP-2 in tissue culture supernatants, serum, plasma, urine, synovial fluid and tissue homogenates. Active MMP-2 may be measured in the range 0.75 to 12 ng/mL and the sensitivity of the assay is 0.5 ng/mL. A longer incubation protocol enables an assay range of 0.19 to 3 ng/mL with a sensitivity of 190 pg/mL, should higher sensitivity be required. The assay uses the pro form of a detection enzyme that can be activated by captured active MMP-2, into an active detection enzyme, through a single proteolytic event. The natural activation sequence in the pro detection enzyme has been replaced using Protein engineering, with an artificial sequence recognised by specific matrix metalloProteinases. MMP activated detection enzyme can then be measured using a specific chromogenic peptide substrate. Standards and samples are incubated in microplate wells precoated with anti-MMP-2 antibody. Any MMP-2 present will be bound to the wells, other components of the sample being removed by washing and aspiration. Either the endogenous levels of free active MMP-2, or the total levels of free MMP-2 in a sample, can be detected. In order to measure the total MMP-2 content, any bound MMP-2 in its pro form is activated using p-aminophenylmercuric acetate (APMA). The standard is pro MMP-2 which is activated in parallel for both types of sample. Active MMP-2 is detected without APMA treatment. The resultant colour is read at 405 nm in a microplate spectrophotometer. The concentration of active MMP-2 in a sample is determined by interpolation from a standard curve. Each pack contains reagents for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate. The Biotrak MMP-2 activity assay has been specifically designed for research purposes.
Matrix Metalloproteinase-2, Biotrak Assay.
Matrix metalloproteinase-2 (MMP-2), also known as gelatinase or type IV collagenase, is a 72 kDa protein. MMP-2 is a member of matrix metalloproteinase (MMP) family of enzymes. The basic structure of MMP2 contains a signal peptide domain that targets the enzyme for secretion, the pro-peptide domain, which is removed when the enzyme is activated and the catalytic site containing gelatin-binding domain.

Biochem/physiol Actions

Matrix metalloproteinase-2 (MMP-2) cleaves gelatin, type IV, V, VII, X, and XI collagens, fibronectin, elastin, laminin, proteoglycans and a range of non-extracellular matrix (ECM) components. Both MMP-2 and MMP9 play an essential role in matrix degradation and they are implicated in the maintenance of neovascularization. MMP-2 activity is associated with human ovarian follicular development. Its expression is elevated in human myocardial infarction and dilated cardiomyopathy. Wound fluid from chronic leg ulcers show elevated expression of MMP2. Overexpression of active MMP-2 affects wound healing in chronic leg ulcers. MMP2 activity is inhibited by tissue inhibitor of metalloproteinase-2 (TIMP-2). Human follicular fluid MMP-2 level acts as a potential biomarker of oocyte maturation in in vitro fertilization and intracytoplasmic sperm injection (ICSI) cycles.

Features and Benefits

  • Specific for MMP-2.
  • Nonradioactive microplate-based assay.
  • Precise, accurate, quantitative measurement.
  • Applicable to complex biological samples.
  • Ready-to-use detection enzyme and substrate.
  • Equivalent sensitivity to ELISA.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak MMP-2 and TIMP-2 Human ELISAs
These products are manufactured by FUJI CHEMICAL INDUSTRIES, Ltd. (Japan).
Biotrak is a trademark of Cytiva

Pictograms

Environment

Signal Word

Danger

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix metalloproteinase 2 level in human follicular fluid is a reliable marker of human oocyte maturation in in vitro fertilization and intracytoplasmic sperm injection cycles.
Yang WJ, et al.
Reproductive Biology and Endocrinology, 13(1), 102-102 (2015)
Immunohistochemical expression of MMP-14 and MMP-2, and MMP-2 activity during human ovarian follicular development.
Vos MC, et al.
Reproductive Biology and Endocrinology, 12(1), 12-12 (2014)
Matrix metalloproteinases, a disintegrin and metalloproteinases, and a disintegrin and metalloproteinases with thrombospondin motifs in non-neoplastic diseases.
Shiomi T
Pathology International, 60(7), 477-496 (2010)
Wound fluid from chronic leg ulcers contains elevated levels of metalloproteinases MMP-2 and MMP-9.
Wysocki AB
The Journal of Investigative Dermatology, 101(1), 64-68 (1993)
Thrombospondin-1 suppresses spontaneous tumor growth and inhibits activation of matrix metalloproteinase-9 and mobilization of vascular endothelial growth factor.
Rodriguez-Manzaneque JC
Proceedings of the National Academy of Sciences of the USA, 98(22), 12485-12490 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service