Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9574-99

TALON® Superflow

Cytiva 28-9574-99, pack of 10 mL

Synonym(s):

TALON Affinity Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

TALON® Superflow Cobalt-IMAC

packaging

pack of 10 mL

manufacturer/tradename

Cytiva 28-9574-99

matrix

6% cross-linked agarose

particle size

60-160 μm

cleaning in place

2-14(Co2+-stripped medium)

working range

3-12(Co2+-stripped medium)

capacity

≥20 mg binding capacity (histidine-tagged protein)(The binding capacity for individual proteins may vary)

storage temp.

2-8°C

Related Categories

General description

Talon® Superflow is a cobalt-based IMAC medium offering different selectivity compared to nickel-charged media.

Features and Benefits

  • Cobalt-based chromatography medium designed for purification of histidine-tagged recombinant Proteins by immobilized metal affinity chromatography (IMAC).
  • Suitable for IMAC purification of histidine-tagged Proteins when Ni2+ is not the optimal choice of metal ion.
  • Compatible with commonly used IMAC reagents and appropriate for purifying Proteins under native or denaturing conditions.
  • Allows for Protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Superflow is a trademark of Sterogene Bioseparations Inc.
Talon is a registered trademark of Clontech

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

His GraviTrap™ TALON® columns are prepacked with 1 ml of TALON® Superflow medium. Each column provides simple manual purification of up to 15 mg of histidine-tagged proteins.

Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.

How to optimize purification of histidine-tagged proteins using Cytiva products.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

See All

Protocols

This page shows how to purify histidine-tagged proteins using TALON Superflow from Cytiva.

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service