Skip to Content
Merck
All Photos(1)

Documents

GE27-4565-01

Salmon Testes Sonicated DNA

Cytiva 27-4565-01, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.52

form

solid

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 27-4565-01

color

white

shipped in

dry ice

storage temp.

−20°C

General description

DNA (Salmon Testes), sonicated
Genomic DNA sheared by sonication for use in hybridization experiments

  • Purified by phenol extraction and freeze dried.
  • Suitable for blocking non-specific binding sites in hybridization experiments to reduce background.
  • Fragmented DNA with a size range from 200-6000 nucleotides.
  • Tested for absence of nuclease.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service