Skip to Content
Merck
All Photos(1)

Key Documents

D7011

Sigma-Aldrich

Deoxyribonucleic acid sodium salt from human placenta

Type XIII

Synonym(s):

hpDNA, DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

type

Type XIII

form

powder

storage temp.

2-8°C

InChI

1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)

InChI key

AWBASQCACWFTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deoxyribonucleic acid (DNA) from placenta and its methylation pattern is indicative of anomalies in the fetal growth.[1] Tobacco smoking during pregnancy favors methylation and epigenetic modulation of placental DNA.[2]

Application

Deoxyribonucleic acid sodium salt from human placenta has been used:
  • as a standard for optimization of digestion followed by high-performance liquid chromatography (HPLC) and mass spectroscopy[3]
  • as a standard for ultra violet (UV) spectrophotometer measurements[4]
  • in polymerase chain reaction (PCR) assay and for standard curve generation for human papillomavirus type 6 quantification[5]

Human placenta DNA (hpDNA) is a natural DNA used in studies of DNA binding anticancer agents and DNA binding agents that modulate DNA structure and function. Human placenta DNA is also used in physicochemical studies of DNA behavior in solution.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA methylation in the human placenta and fetal growth
Koukoura O, et al.
Molecular Medicine Reports, 5(4), 883-889 (2012)
Johan S Nilsson et al.
Scientific reports, 9(1), 15432-15432 (2019-10-30)
Nasopharyngeal cancer (NPC) is associated with the Epstein-Barr virus (EBV). The clinical presentation and prognosis of NPC is well described, but not in relation to intralesional EBV-DNA load. In a retrospective design, 48 patients with NPC were examined. Patient history
Results from the NIST 2004 DNA quantitation study
Kline MC, et al.
Journal of Forensic Sciences, 50(3), 1-8 (2005)
Viral load and m RNA expression of HPV type 6 among cases with recurrent respiratory papillomatosis
Forslund O, et al.
Laryngoscope, 126(1), 122-127 (2016)
Genome-wide DNA methylation study in human placenta identifies novel loci associated with maternal smoking during pregnancy
Morales E, et al.
International Journal of Epidemiology, 45(5), 1644-1655 (2016)

Questions

  1. This item (D7011) has been discontinued. Can you please let me know what the comparable product is?

    1 answer
    1. Product D3035, is a solution of Deoxyribonucleic acid prepared from human placenta at a concentration of 0.05 mg/mL (the 0.1 mg pack size would be 2 mL) and would be the closest equivalent alternative to the discontinued D7011. Please see the link below to review this product:
      https://www.sigmaaldrich.com/product/sigma/d3035

      If a powdered form is preferred we do offer comparable alternatives such as 11691112001 from Roche, Human Genomic DNA from human blood (buffy coat). It is not from human placenta but prepared from 100 to 200 individual blood donors of both sexes (pool of the buffy coat). If the DNA from another species is compatible in your application, products D1626 (from salmon) or D6898 (from herring) are options to consider.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service