GE17-5468-55
HiTrap® Capto™ SP ImpRes
Cytiva 17-5468-55, pack of 5 × 5 mL
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 17-5468-55
parameter
42 psi
bed size
7 mm × 25 mm
bed volume
1 mL
General description
HiTrap® Capto™ SP ImpRes is a strong cation exchanger prepacked with BioProcess Capto™ ImpRes media for screening and small-scale protein purifications using ion exchange chromatography (IEX).
Application
Capto™ SP ImpRes has a strong sulfonate cation exchanger coupled to a chemically modified, high-fLow Agarose matrix. The modified Agarose matrix provides greater particle rigidity without compromising pore size, which allows more flexibility in process-scale applications.
Features and Benefits
- High-throughput medium for intermediate and polishing Protein purification, excellent choice for process-scale purification.
- The combined properties of high binding capacity, high flow rate, and Low back pressure reduces process cycle times and improves productivity at process scale.
- Rigid Agarose matrix enables high bed heights and purification of viscous samples at high flow rates.
- Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
- Strong sulfonate anion exchanger
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Other Notes
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
Legal Information
Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page covers detailed information on cleaning procedures and recommended flow for column cleaning.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service