Skip to Content
Merck
All Photos(1)

Documents

GE27-3565-01

Microspin Columns

Cytiva 27-3565-01, pack of 100 pieces

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

packaging

pack of 100 pieces

manufacturer/tradename

Cytiva 27-3565-01

Related Categories

General description

MicroSpin Columns.

Application

MicroSpin columns are designed for the rapid purification of DNA in conjunction with Sephadex® G-25 DNA Grade for use in a wide range of applications, including desalting, buffer exchange, and removal of unincorporated nucleotides from end-labelled oligonucleotides. They can be used for any DNA greater than 10 bases in length and are therefore highly suitable for the purification of oligonucleotides or very small DNA fragments following synthesis or a labeling reaction.

Features and Benefits

  • For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 to 100 μL using spin-column chromatography and Sephadex® G-25 DNA Grade (not included).
  • Excellent for rapid purification of newly synthesized oligonucleotides > 10-mers in 100 to 150 ?l of deprotection solution using spin-column chromatography.
  • Flexible for your experiments, allowing differing amounts of Sephadex.
  • Can also be used for desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labeled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 μL.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

MicroSpin is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

DNA cleanup using illustra™ GFX kits from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service