Skip to Content
Merck
All Photos(1)

Documents

GE17-0446-01

Calibration Kit Low Molecular Weight For Electrophoresis

Cytiva 17-0446-01

Synonym(s):

Calibration Kit, Low Molecular Weight Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.32

form

solid

feature

wetted part: no

packaging

pkg of 10 vials

manufacturer/tradename

Cytiva 17-0446-01

storage condition

(Unopened vials should be s-red at 4°C)

color

White

storage temp.

2-8°C

Related Categories

General description

The calibration kit for low molecular weight is specifically designed for electrophoresis applications, providing precise standards for accurate molecular weight determination.

Application

For MW determinations by polyacrylamide gel electrophoresis (PAGE) in homogeneous or gradient gels.

Features and Benefits

  • Determine molecular weight of uncharacterized protein samples with precision by comparison with carefully selected standards.
  • Get clear, sharp bands for easy measurement of migration distances.
  • Bands can be visualized by using Deep Purple Total Protein Stain, Coomassie Brilliant Blue, or silver staining.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service