Skip to Content
Merck
All Photos(1)

Key Documents

M3913

Supelco

SigmaMarker

low range, mol wt 6,500-66,000 Da

Synonym(s):

protein markers, protein standards

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$95.90
10 VIALS
$466.00

$95.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 VIAL
$95.90
10 VIALS
$466.00

About This Item

MDL number:
UNSPSC Code:
41105337
NACRES:
NA.25

$95.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

form

lyophilized powder

usage

sufficient for 10-20 large gels (16 × 18 cm)
sufficient for 20-30 mini-gels (10 × 10 cm)
 vial sufficient for 50-100 PhastGels

mol wt

6,500-66,000 Da

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

SigmaMarkers are designed for use with Laemmli SDS-PAGE, the PhastSystem (Pharmacia) and Schagger and von Jagow SDS-PAGE (Tricine). SigmaMarkers cover the range of molecular weights common to most proteins or their subunits. The wide, high and low range markers are lyophilized with sample buffer so that they are ready-to-use after reconstitution with deionized water. The markers are formulated to yield a distribution of well-defined bands of approximately equal intensity after electrophoresis and Coomassie blue staining.

Other Notes

Protein (mol. wt.)
Aprotinin, bovine lung (6,500)
α-Lactalbumin, bovine milk (14,200)
Trypsin inhibitor, soybean (20,000)
Trypsinogen, bovine pancreas (24,000)
Carbonic anhydrase, bovine erythrocytes (29,000)
Glyceraldehyde-3-phosphate dehydrogenase, rabbit muscle (36,000)
Ovalbumin, chicken egg (45,000)
Albumin, bovine serum (66,000)
Will yield eight protein bands with Coomassie Blue staining.

Reconstitution

Reconstitution with 100 μl of deionized water results in a solution containing 2-3.5 mg of protein per ml of 62 mM Tris-HCl, pH 6.8, 1 mM EDTA, 4% sucrose, 10 mM TCEP, 2% SDS, and 0.01% bromophenol blue.

Legal Information

SigmaMarker is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Jun Lee et al.
Applied and environmental microbiology, 68(8), 3919-3924 (2002-07-31)
Gram-negative bacteria can communicate with each other by N-acyl homoserine lactones (AHLs), which are quorum-sensing autoinducers. Recently, the aiiA gene (encoding an enzyme catalyzing the degradation of AHL) has been cloned from Bacillus sp. strain 240B1. During investigations in the
Andrés González et al.
Biochemistry and molecular biology education : a bimonthly publication of the International Union of Biochemistry and Molecular Biology, 46(5), 493-501 (2018-08-02)
Recombinant protein expression and site-directed mutagenesis of target genes have demonstrated an increasing importance in the fields of molecular biology, biochemistry, biotechnology, and medicine. By using the flavodoxin of the model cyanobacterium Anabaena sp. PCC 7120 as a laboratory tool
Antonios Matsakas et al.
Experimental physiology, 91(6), 983-994 (2006-07-29)
The aim of this study was to test the hypothesis that swimming training might impact differentially myostatin expression in skeletal muscles, depending on fibre type composition, and in cardiac muscle of rats. Myostatin expression was analysed by real time reverse
Shalini Iyer et al.
PeerJ, 6, e5815-e5815 (2018-10-26)
Amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD), two late onset neurodegenerative diseases, have been shown to share overlapping cellular pathologies and genetic origins. Studies suggest that a hexanucleotide repeat expansion in the first intron of the C9orf72 gene is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service