Key Documents
GE10600102
Amersham Hybond® LFP Western blotting membranes, PVDF
pore size 0.2 μm, sheet W × L 80 mm × 90 mm, pkg of 25 ea
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva 10600102
sheet W × L
80 mm × 90 mm
pore size
0.2 μm pore size
General description
Low-fluorescent, hydrophobic PVDF protein blotting membrane with 0.2 µm pore size. Amersham Hybond LFP 0.2 PVDF is a low-fluorescent, hydrophobic membrane, with 0.2 µm pore size, for protein transfer. Amersham Hybond LFP is optimized for fluorescent detection in Western blotting applications and for use with the Amersham ECL Plex detection system. This membrane has low background fluorescence, resulting in high sensitivity. Typical protein binding capacity is > 200 µg/cm2. The PVDF protein blotting membrane, with 0.2 µm pore size, provides low auto-fluorescence across a wide range of excitation/emission wavelengths.
Features and Benefits
  • Much lower background fluorescence than other commercially available PVDF membranes, resulting in higher sensitivity.
  • Protein binding capacity over 200 µg/cm2.
  • Compatible with a variety of blocking agents, transfer buffers, and total protein blot stains.
  • Available in popular precut sizes for laboratory applications.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)